Skip to Content
Merck
All Photos(2)

Key Documents

908509

Sigma-Aldrich

Y6

Synonym(s):

BTP-4F, Non fullerene acceptor Y6, PCE 157

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$465.00

$465.00


Estimated to ship onMarch 19, 2025


Request a Bulk Order

Select a Size

Change View
100 MG
$465.00

About This Item

Empirical Formula (Hill Notation):
C82H86F4N8O2S5
CAS Number:
Molecular Weight:
1451.93
MDL number:
UNSPSC Code:
32111701
NACRES:
NA.23

$465.00


Estimated to ship onMarch 19, 2025


Request a Bulk Order

description

Band gap: 1.55 eV

Assay

≥99%

form

solid

solubility

chloroform: soluble

Orbital energy

HOMO -5.65 eV 
LUMO -4.10 eV 

SMILES string

Fc1cc2c(cc1F)C(=C(C#N)C#N)\C(=C\c3[s]c4c([s]c5c4[n](c6c7[n](c8c([s]c%10c8[s]c(c%10CCCCCCCCCCC)\C=C%11/C(=O)c%12c(cc(c(c%12)F)F)C/%11=C(C#N)C#N)c7c9n[s]nc9c65)CC(CCCC)CC)CC(CCCC)CC)c3CCCCCCCCCCC)\C2=O

InChI key

XJRVXAOYOOOQLU-LAGONYLDSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Non-fullerene acceptors (NFAs) are currently a major focus of research in the development of bulk-heterojunction organic solar cells (OSCs). In contrast to the widely used fullerene acceptors (FAs), the optical properties and electronic energy levels of NFAs can be designed and readily tuned. NFA-based OSCs can also achieve greater thermal stability and photochemical stability, as well as longer device lifetimes, than their FA-based counterparts.Recent developments have led to a rapid increase in power conversion efficiencies for NFA OSCs, with values now exceeding 15% in a single junction cell, and >17% for a tandem cell, demonstrating the viability of using NFAs to replace FAs in next-generation high-performance OSCs.
Y6 is a non-fullerene acceptor-donor-acceptor (A-D-A) type small molecular acceptor (SMA) with flexible alkyl chains and a centrally fused ring. It has a ladder-type electron-deficient core, which can be blended with PM6 for organic photovoltaic applications.[1][2][3]

Application

Y6 is a non-fullerene acceptor (NFA) that uses an electron-deficient molecular core to obtain a low bandgap with improved electron affinity. OPV devices made from Y6 can reach exceptionally high performances in single-junction devices, with a maximum PCE reported of 15.7% (14.9% certified by Enli Tech Laboratory) when paired with PM6. Y6 is versatile and can be employed in both conventional and inverted OPV device architectures and can maintain a high PCE with varying active layer thicknesses (13.6% in thick 300 nm layers).
Y6 is used as an electron acceptor material in organic solar cells. It has shown promising results in improving the power conversion efficiency of organic solar cells. Y6 and its derivatives are also used in material engineering studies to understand the relationships between molecular structure, morphology and device performance in organic photovoltaics.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Fine-Tuning Energy Levels via Asymmetric End Groups Enables Polymer Solar Cells with Efficiencies over 17\%
Luo Z, et al.
Joule, 31(39), 1903441-1903441 (2020)
Eco-compatible solvent-processed organic photovoltaic cells with over 16% efficiency
Hong L, et al.
Advanced Materials, 31(39), 1903441-1903441 (2019)
Single-Junction Organic Solar Cell with over 15% Efficiency Using Fused-Ring Acceptor with Electron-Deficient Core
Yuan J, et al.
Joule, 3(4), 1140-1151 (2019)
PTPsigma inhibitors promote hematopoietic stem cell regeneration
Zhang Y, et al.
Nature Communications, 10(1), 1-15 (2019)

Articles

To achieve net-zero emissions by 2050, renewable power contributions must triple. Photovoltaic stations provide vital utility power, achieved primarily through third- and fourth-generation technology. Promising trends include recycling and revolutionary, ultra-lightweight, flexible, and printable solar cells.

Professor Chen (Nankai University, China) and his team explain the strategies behind their recent record-breaking organic solar cells, reaching a power conversion efficiency of 17.3%.

Related Content

Organic electronics utilizes organic conductors and semiconductors for applications in organic photovoltaics, organic light-emitting diodes, and organic field-effect transistors.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service