Skip to Content
Merck
All Photos(1)

Key Documents

933678

Sigma-Aldrich

Y6(BTPTT-4F)

Synonym(s):

Y6, 2,2′-((2Z,2′Z)-((12,13-Bis(2-ethylhexyl)-3,9-diundecyl-12,13-dihydro-[1,2,5]thiadiazolo[3,4-e]thieno-[2",3":4′,5′]thieno[2′,3′:4,5]pyrrolo[3,2-g]thieno-[2′,3′:4,5]thieno[3,2-b]indole-2,10-diyl)bis(methanylylidene))-bis(5,6-difluoro-3-oxo-2,3-dihydro-1H-indene-2,1-diylidene))dimalononitrile

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C82H86F4N8O2S5
CAS Number:
Molecular Weight:
1451.93
MDL number:
UNSPSC Code:
32111701
NACRES:
NA.21
Pricing and availability is not currently available.

Assay

≥99% (H-NMR)

Quality Level

color

black

solubility

chloroform: soluble

λmax

731 nm in chloroform (UV)

Orbital energy

HOMO - 5.65 eV 
LUMO - 4.1 eV 

SMILES string

Fc1cc2c(cc1F)C(=C(C#N)C#N)\C(=C\c3[s]c4c([s]c5c4[n](c6c7[n](c8c([s]c%10c8[s]c(c%10CCCCCCCCCCC)\C=C%11/C(=O)c%12c(cc(c(c%12)F)F)C/%11=C(C#N)C#N)c7c9n[s]nc9c65)CC(CCCC)CC)CC(CCCC)CC)c3CCCCCCCCCCC)\C2=O

InChI

1S/C82H86F4N8O2S5/c1-7-13-17-19-21-23-25-27-29-33-51-63(39-57-65(49(41-87)42-88)53-35-59(83)61(85)37-55(53)75(57)95)97-81-73-79(99-77(51)81)67-69-70(92-101-91-69)68-72(71(67)93(73)45-47(11-5)31-15-9-3)94(46-48(12-6)32-16-10-4)74-80(68)100-78-52(34-30-28-2

InChI key

XJRVXAOYOOOQLU-LAGONYLDSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Y6 (BTP-4F) is a popular non-fullerene acceptor (NFA) molecule for use in Organic Photovoltaic (OPV) given its contribution to device power conversion efficiencies (PCEs). Also known BTP-4F, Y6 is a highly conjugated electron deficient organic semiconductor with an A-DAD-A structure. The absorption spectrum of Y6 molecule has a maximum at around 810 nm and extends to 1100 nm. This means that Y6 and its polymer blends have the potential to absorb light across the entire visible and near infra-red spectrum.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Edgar Gutierrez-Fernandez et al.
Advanced science (Weinheim, Baden-Wurttemberg, Germany), 9(1), e2104977-e2104977 (2021-12-03)
There is a growing demand to attain organic materials with high electron mobility, μe , as current reliable reported values are significantly lower than those exhibited by their hole mobility counterparts. Here, it is shown that a well-known nonfullerene-acceptor commonly
Triplet-Charge Annihilation in a Small Molecule Donor: Acceptor Blend as a Major Loss Mechanism in Organic Photovoltaics
J. Marin-Beloqui et al.
Advanced Energy Materials, 11, 2100539-2100539 (2021)
From Y6 to BTPT-4F: a theoretical insight into the influence of the individual change of fused-ring skeleton length or side alkyl chains on molecular arrangements and electron mobility.
Zhang, Jie, et al.
New. J. Chem., 45, 12247?12259-12247?12259 (2021)
You Chen et al.
Chemical communications (Cambridge, England), 55(47), 6708-6710 (2019-05-22)
For photovoltaic polymers with a D-π-A backbone, there are a great deal of D and A units, but the choice of π bridge is relatively limited and thiophene (T) is still the most effective one. Here, we utilize two D-π-A
Over 17% efficiency ternary organic solar cells enabled by two non-fullerene acceptors working in an alloy-like model.
Zhan, Lingling et al.
Energy & Environmental Science, 13, 635-645 (2020)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service