Skip to Content
Merck
All Photos(1)

Key Documents

900662

Sigma-Aldrich

Poly(D,L-lactide-co-glycolide)(85/15)-b-poly(ethylene glycol)-carboxylic acid

5k-13k

Synonym(s):

PLGA-PEG, PLGA-PEG Carboxylic acid diblock copolymer, PLGA-PEG-COOH, PLGA-b-PEG

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
$270.00

$270.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
250 MG
$270.00

About This Item

Linear Formula:
HO(CH(CH3)COO)x(CH2COO)y(CH2CH2O)nH
UNSPSC Code:
12352106
NACRES:
NA.23

$270.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

form

solid

storage temp.

2-8°C

Application

Biocompatible, amphiphilic block copolymer composed of a hydrophilic PEG block and a hydrophobic Poly(lactide-co-glycolide) (PLGA) block, functionalized with a terminal carboxylic acid. These materials have been used in control release and nanoparticle formulation for drug delivery applications. Well-defined materials with varying properties can be prepared by controlling the relative length of each polymer block. Additionally, the ratio of lactide to glycolide can be tuned to control rate of degradation. Carboxylic acid termination allows for facile further chemical modification of these materials.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Miles A Miller et al.
Nature communications, 6, 8692-8692 (2015-10-28)
Therapeutic nanoparticles (TNPs) aim to deliver drugs more safely and effectively to cancers, yet clinical results have been unpredictable owing to limited in vivo understanding. Here we use single-cell imaging of intratumoral TNP pharmacokinetics and pharmacodynamics to better comprehend their
Jijin Gu et al.
Molecular pharmaceutics, 12(8), 2889-2903 (2015-06-24)
The goal of this study was to develop and characterize a novel intravaginal film platform for targeted delivery of small interfering RNA (siRNA)-loaded nanoparticles (NP) to dendritic cells as a potential gene therapy for the prevention of sexually transmitted human
Soroush Ardekani et al.
Scientific reports, 5, 16258-16258 (2015-11-21)
Nitroglycerin (NTG) markedly enhances nitric oxide (NO) bioavailability. However, its ability to mimic the anti-inflammatory properties of NO remains unknown. Here, we examined whether NTG can suppress endothelial cell (EC) activation during inflammation and developed NTG nanoformulation to simultaneously amplify

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service