Skip to Content
Merck
All Photos(1)

Key Documents

719897

Sigma-Aldrich

Resomer® RG 502 H, Poly(D,L-lactide-co-glycolide)

acid terminated, viscosity 0.16-0.24 dL/g 

Synonym(s):

PLGA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[C3H4O2]x[C2H2O2]y
CAS Number:
MDL number:
UNSPSC Code:
12162002
NACRES:
NA.23

Quality Level

form

amorphous

feed ratio

lactide:glycolide 50:50

degradation timeframe

<3 months

viscosity

0.16-0.24 dL/g

transition temp

Tg 42-46 °C

storage temp.

2-8°C

SMILES string

O2C(C(=O)OC(C2=O)C)C.O1CC(=O)OCC1=O

InChI

1S/C6H8O4.C4H4O4/c1-3-5(7)10-4(2)6(8)9-3;5-3-1-7-4(6)2-8-3/h3-4H,1-2H3;1-2H2

InChI key

LCSKNASZPVZHEG-UHFFFAOYSA-N

Application

Poly(D,L-lactide-co-glycolide) (PLGA) canbe used:

  • In the formulation ofa non-aqueous drug delivery system for H2S donors to aid thesustained release of the drug.
  • To prepare a polymer blend containing Ganciclovir-loaded microspheresdispersed in PLGA–PEG–PLGA polymer gel. This formulation can be used as asustained delivery system for Ganciclovir drug.
It can also be used to fabricate NO-releasingglucose and lactate sensors.

Legal Information

Product of Evonik
RESOMER is a registered trademark of Evonik Rohm GmbH

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Patil et al.
AAPS PharmSciTech, 18(6), 2291-2302 (2017-01-20)
Hydrogen sulfide (H2S) targets both underlying factors in glaucoma pathogenesis by reducing elevated intraocular pressure (IOP) and providing retinal neuroprotection, whereas the current clinical approaches targets only reducing IOP. Therefore, H2S could be a potential superior candidate for glaucoma pharmacotherapy.
Diazeniumdiolate-doped poly (lactic-co-glycolic acid)-based nitric oxide releasing films as antibiofilm coatings.
Cai W,et al.
Biomaterials, 33(32), 7933-7944 (2012)
Sridhar Duvvuri et al.
Journal of controlled release : official journal of the Controlled Release Society, 108(2-3), 282-293 (2005-10-19)
The purpose of this work is to develop empirical equations for describing the in vitro ganciclovir (GCV) release from PLGA microspheres and also to develop and characterize a formulation containing GCV loaded PLGA microspheres dispersed in thermogelling PLGA-PEG-PLGA polymer gel.
Qinyi Yan et al.
Biosensors & bioelectronics, 26(11), 4276-4282 (2011-05-20)
Intravenous amperometric needle-type enzymatic glucose/lactate sensors intended for continuous monitoring are prepared with a novel nitric oxide (NO) releasing layer to improve device hemocompatibility. To create an underlying NO release coating, the sensors with immobilized enzymes (either glucose oxidase or
Z Zhuang et al.
Journal of dental research, 98(2), 200-208 (2018-11-06)
Periodontitis is characterized by the progressive destruction of tooth-supporting alveolar bone, which is mainly caused by chronic inflammation in response to persistent bacterial insult. It has recently become clear that the pathogenesis of periodontitis is associated with a high ratio

Articles

Interest in utilizing biodegradable polymers for biomedical applications has grown since the 1960s.

Synthetic aliphatic polyesters dominate resorbable biomaterials in clinical use.

AliAliphatic polyesters, including polylactide and polyglycolide, are biodegradable polymers widely used in medical applications.

Immunosuppressive tumor-associated myeloid cells (TAMC) are responsible for glioblastoma (GBM) resistance to immunotherapies and existing standard of care treatments. This mini-review highlights recent progress in implementing nanotechnology in advancing TAMC-targeted therapies for GBM.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service