Skip to Content
Merck
All Photos(2)

Key Documents

430471

Sigma-Aldrich

Poly(D,L-lactide-co-glycolide)

ester terminated, Mw 50,000-75,000

Synonym(s):

Lactel® B6006-1, PLGA

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$102.00
5 G
$351.00

$102.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$102.00
5 G
$351.00

About This Item

Linear Formula:
[C3H4O2]x[C2H2O2]y
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

$102.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

form

amorphous

feed ratio

lactide:glycolide 85:15

mol wt

Mw 50,000-75,000

degradation timeframe

<6 months

viscosity

0.55-0.75 dL/g, 0.1 % (w/v) in chloroform(25 °C)

transition temp

Tg 45-50 °C

solubility

ethyl acetate, chloroform, acetone and THF: soluble

storage temp.

2-8°C

SMILES string

OCC(O)=O.CC(O)C(O)=O

InChI

1S/C3H6O3.C2H4O3/c1-2(4)3(5)6;3-1-2(4)5/h2,4H,1H3,(H,5,6);3H,1H2,(H,4,5)

InChI key

XBBVURRQGJPTHH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Characteristic properties of ester terminated poly(D,L-lactide-co-glycolide) (PLGA) are in vivo biodegradability, tunable drug release profile and very high encapsulation capacity.[1]

Application

PLGA polymers find uses in the production of biodegradable medical sutures and for controlled drug release. PLGA microspheres have been reported for use in controlled release of human growth hormone.[1] Thin membranes of blended polymers constituting of PLGA/polycaprolactone/β-tricalcium phosphate have been used in guided bone regeneration.[2]

Features and Benefits

Controlled release of bioactive agents, sutures and bioabsorbable implantable devices.

Physical form

Biocompatible, biodegradable polymer.

Legal Information

Lactel is a registered trademark of Evonik Corp

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fabrication of blended polycaprolactone/poly (lactic-co-glycolic acid)/?-tricalcium phosphate thin membrane using solid freeform fabrication technology for guided bone regeneration
Shim JH, et al.
Tissue Engineering: Part A, 19(3-4), 317-328 null
Preparation and characterization of poly (D, L-lactide-co-glycolide) microspheres for controlled release of human growth hormone
Capan Y, et al.
Aaps Pharmscitech, 4(2), 147-156 (2003)
Manjae Gil et al.
Advanced science (Weinheim, Baden-Wurttemberg, Germany), 5(6), 1800024-1800024 (2018-06-26)
Anisotropically compartmentalized microparticles have attracted increasing interest in areas ranging from sensing, drug delivery, and catalysis to microactuators. Herein, a facile method is reported for the preparation of helically decorated microbuilding blocks, using a modified electrohydrodynamic cojetting method. Bicompartmental microfibers
Ayelen L Helling et al.
Tissue engineering. Part A, 25(3-4), 214-223 (2018-10-04)
In human skin the junction between the epidermis and dermis undulates. Epidermal stem cells pattern according to their position relative to those undulations. Here we describe a rig in which epidermal cells are cultured on a collagen-coated poly(d,l-lactide-co-glycolide) (PLGA) membrane.
Teresa Musumeci et al.
International journal of pharmaceutics, 440(2), 135-140 (2012-10-20)
Melatonin, a neurohormone secreted by the pineal gland, is able to modulate intraocular pressure (IOP). The aim of this study was to generate nanoparticle (NPs) sustained release formulations that allow to extend the pre-corneal residence time of melatonin, thus prolonging

Articles

Interest in utilizing biodegradable polymers for biomedical applications has grown since the 1960s.

Synthetic aliphatic polyesters dominate resorbable biomaterials in clinical use.

AliAliphatic polyesters, including polylactide and polyglycolide, are biodegradable polymers widely used in medical applications.

Immunosuppressive tumor-associated myeloid cells (TAMC) are responsible for glioblastoma (GBM) resistance to immunotherapies and existing standard of care treatments. This mini-review highlights recent progress in implementing nanotechnology in advancing TAMC-targeted therapies for GBM.

See All

Questions

1–8 of 8 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  3. What is the tensile strength of Product No. 403471 Poly(DL-lactide-co-glycolide)?

    1 answer
    1. Product 430471 has a tensile strength of 6000 - 8000 psi, with an elongation of 3-10%.

      Helpful?

  4. Is Product No. 403471 a resorbable polymer?

    1 answer
    1. Yes, Product No. 430471 has a resorption time of 5-6 months.

      Helpful?

  5. What is the density of Product No. 430471 Poly(DL-lactide-co-glycolide)?

    1 answer
    1. According to the manufacturer, product 430471 has a theoretical density of 1.27g/mL.

      Helpful?

  6. What are the end groups of Product No. 430471 (Poly(DL-lactide-co-glycolide)?

    1 answer
    1. Product 430471 has a dodecyl (12 carbon) ester on one end of the polymer chain and a hydroxyl end group on the other.

      Helpful?

  7. How is this Product No. 430471 synthesized (Poly(DL-lactide-co-glycolide)?

    1 answer
    1. To synthesize product 430471, 1-dodecanol is used as an initiator and stannous octoate is used as a catalyst to start the ring opening polymerization of D,L-lactide and glycolide monomers.

      Helpful?

  8. What is the Department of Transportation shipping information for this product?

    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service