Skip to Content
Merck
All Photos(1)

Documents

937754

Sigma-Aldrich

Poly(D,L-lactic-co-glycolic acid)-block-poly(glutamic acid)

PLGA average Mn 25,000, mol wt (PGlu average Mn 5,000), lactide:glycolide 50:50

Synonym(s):

Amphiphilic, Block polymer, Drug delivery, Glutamic acid, PLGA, PLGA-b-PGlu, Poly(glutamic acid), Polyaminoacids

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[(C3H4O2)a(C2H2O2)b]n O[C5H7NO3]m
UNSPSC Code:
12162002
NACRES:
NA.21

feed ratio

lactide:glycolide 50:50

Quality Level

mol wt

PLGA average Mn 25,000
(PGlu average Mn 5,000)

color

white to beige, powder

storage temp.

−20°C

Application

This polymer is a amphiphilic diblock copolymer composed of a hydrophobic PLGA block and a hydrophilic poly(glutamic acid) block. PLGA is a biocompatible and biodegradable polymer that has been approved by the FDA and the EMA for biomedical and pharmaceutical applications. Poly(glutamic acid) is biodegradable, non-toxic and non-immunogenic macromolecules. This biodegradable, biocompatible polymers can self-assemble to form nanoparticles, such as micelles and polymersomes, for drug delivery applications

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ibrahim R Khalil et al.
International journal of molecular sciences, 18(2) (2017-02-06)
In the past decade, poly-γ-glutamic acid (γ-PGA)-based micro/nanoparticles have garnered remarkable attention as antimicrobial agents and for drug delivery, owing to their controlled and sustained-release properties, low toxicity, as well as biocompatibility with tissue and cells. γ-PGA is a naturally
Yung-Chih Kuo et al.
Colloids and surfaces. B, Biointerfaces, 88(1), 158-164 (2011-07-19)
Nanoparticles (NPs) with ternary components of polyethyleneimine (PEI), poly-(γ-glutamic acid) (γ-PGA), and poly(lactide-co-glycolide) (PLGA) were applied to carry and release saquinavir (SQV). Hydrophobic SQV was encapsulated in the particle core composed of PLGA to form SQV-PLGA NPs, and the surface

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service