Skip to Content
Merck
All Photos(2)

Key Documents

805726

Sigma-Aldrich

Poly(D,L-lactide-co-glycolide)

lactide:glycolide 50:50, Mw 45,000 g/mol

Synonym(s):

PLGA

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$514.00

$514.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$514.00

About This Item

Linear Formula:
[C3H4O2]x[C2H2O2]y
MDL number:
UNSPSC Code:
12162002
NACRES:
NA.23

$514.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

description

degradation timeframe: <3 months

Quality Level

form

crystals

mol wt

Mw 45,000 g/mol

composition

lactide:glycolide, 50:50

storage temp.

2-8°C

SMILES string

OCC(O)=O.CC(O)C(O)=O

InChI

1S/C3H6O3.C2H4O3/c1-2(4)3(5)6;3-1-2(4)5/h2,4H,1H3,(H,5,6);3H,1H2,(H,4,5)

InChI key

XBBVURRQGJPTHH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Polymer used to make controlled release nanoparticles or scaffolds for biomedical applications.

Features and Benefits

Controlled release of bioactive agents, sutures and bioabsorbable implantable devices.

Physical form

Biocompatible, biodegradable polymer.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service