Skip to Content
Merck
All Photos(2)

Key Documents

901021

Sigma-Aldrich

Poly(L-lactide-co-glycolide)

greener alternative

10:90, viscosity 1.7 dL/g 

Synonym(s):

PLGA

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$451.00

$451.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$451.00

About This Item

Linear Formula:
(C4H4O4)m(C6H8O4)n
UNSPSC Code:
12162002
NACRES:
NA.25

$451.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

form

chips

feed ratio

lactide:glycolide 10:90±5

greener alternative product characteristics

Design for Energy Efficiency
Use of Renewable Feedstocks
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

color

light yellow to tan

viscosity

1.4-2.0 dL/g
1.7 dL/g

greener alternative category

shipped in

dry ice

storage temp.

−20°C

General description

Poly(L-lactide-co-glycolide) (PLGA) is a biodegradable and biocompatible copolymer synthesized from the ring opening polymerization of lactide and glycolide. Due to the ester linkages found in the polymer backbone, these polymers are able to degrade via hydrolysis. Depending on the ratio of monomers used and polymer block structure, different forms of PLGA can be obtained. These differing forms yield materials with different degrees of crystallinity, thermal transitions, solubility, and rates of degradation.
We are committed to bringing you Greener Alternative Products, which belongs to one of the four categories of greener alternatives. This is an enabling product that belongs to a family of high-performance bio-based polymers that are essential for developing efficient, low-cost, and environmentally friendly renewable energy solutions. Click here for more information.

Application

Poly(L-lactide-co-glycolide) (PLGA) is a biodegradable and biocompatible copolymer for use in biomedical research devices, such as grafts, sutures, implants, prosthetic devices, surgical sealant films, scaffolds for tissue engineering applications, micro and nanoparticles. PLGA is also utilized in drug delivery research, such as in the synthesis and application of polymer-drug conjugates.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service