Skip to Content
Merck
All Photos(2)

Key Documents

804975

Sigma-Aldrich

DTBPF-Pd-G3

Synonym(s):

1,1′-Bis(di-tert-butylphosphino)ferrocene Pd G3

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
$118.00
1 G
$399.00

$118.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
250 MG
$118.00
1 G
$399.00

About This Item

Empirical Formula (Hill Notation):
C39H57FeNO3P2PdS
Molecular Weight:
844.15
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

$118.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

form

solid

Quality Level

feature

generation 3

reaction suitability

core: palladium
reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Cross Couplings
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: catalyst

impurities

may contain 10-20% Di-μ-mesylbis[2′-(amino-N)[1,1′-biphenyl]-2-yl-C]dipalladium(II) (Aldrich Number 762156)
may contain 5-10% DTBPF (Aldrich Number 695149)
may contain 5-10% pentane

mp

87-150 °C (decomposition)

functional group

phosphine

InChI

1S/2C13H22P.C12H10N.CH4O3S.Fe.Pd/c2*1-12(2,3)14(13(4,5)6)11-9-7-8-10-11;13-12-9-5-4-8-11(12)10-6-2-1-3-7-10;1-5(2,3)4;;/h2*7-10H,1-6H3;1-6,8-9H,13H2;1H3,(H,2,3,4);;/q;;;;;+1/p-1

InChI key

AGXLTHQKOWPGBX-UHFFFAOYSA-M

General description

DTBPF-Pd-G-3 is a versatile palladium precatalyst developed by Buchwald group. It is widely employed in various cross-coupling reactions involving the formation of C-C, C-N, C-O, C-F, C-CF3, and C-S bonds. Precatalysts effectively generate active catalytic species for use in various reactions.

Application

Pd precatalyst for cross-coupling. Buchwald Third Generation Palladium Precatalyst.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

G3 and G4 Buchwald palladium precatalysts are the newest air, moisture, and thermally stable crossing-coupling complexes used in bond formation for their versatility and high reactivity.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service