Skip to Content
Merck
All Photos(3)

Key Documents

773905

Sigma-Aldrich

RockPhos Pd G3

Synonym(s):

RockPhos-G3-palladacycle, [(2-Di-tert-butylphosphino-3-methoxy-6-methyl-2′,4′,6′-triisopropyl-1,1′-biphenyl)-2-(2-aminobiphenyl)]palladium(II) methanesulfonate

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$151.00
500 MG
$717.00
5 G
$5,320.00

$151.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onMay 30, 2025


Request a Bulk Order

Select a Size

Change View
100 MG
$151.00
500 MG
$717.00
5 G
$5,320.00

About This Item

Empirical Formula (Hill Notation):
C44H62NO4PPdS
CAS Number:
Molecular Weight:
838.43
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

$151.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onMay 30, 2025


Request a Bulk Order

form

solid

Quality Level

feature

generation 3

reaction suitability

core: palladium
reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: catalyst
reaction type: Cross Couplings

mp

220-250 °C

functional group

phosphine

SMILES string

CC1=CC=C(OC)C(P(C(C)(C)C)C(C)(C)C)=C1C2=C(C(C)C)C=C(C(C)C)C=C2C(C)C.NC3=CC=CC=C3C4=CC=CC=C4[Pd]OS(C)(=O)=O

InChI

1S/C31H49OP.C12H10N.CH4O3S.Pd/c1-19(2)23-17-24(20(3)4)28(25(18-23)21(5)6)27-22(7)15-16-26(32-14)29(27)33(30(8,9)10)31(11,12)13;13-12-9-5-4-8-11(12)10-6-2-1-3-7-10;1-5(2,3)4;/h15-21H,1-14H3;1-6,8-9H,13H2;1H3,(H,2,3,4);/q;;;+1/p-1

InChI key

PANIJJUVUGSZSP-UHFFFAOYSA-M

Application

Buchwald 3rd Generation Pd Precatalyst containing bulk RockPhos ligand. This system has been shown to be effective for cross-coupling reactions like the arylation of aliphatic alcohols.


Synthesis and Application of Palladium Precatalysts that Accommodate Extremely Bulky Di-tert-butylphosphino Biaryl Ligands

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

G3 and G4 Buchwald palladium precatalysts are the newest air, moisture, and thermally stable crossing-coupling complexes used in bond formation for their versatility and high reactivity.

Related Content

Explore reliable, premium grade catalysis materials for your pharma or industrial project. Specialty chemicals and formulations are available in bulk quantities and volumes from a few grams to multi-metric tons with complete documentation to simplify your leap from development to commercialization.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service