Skip to Content
Merck
All Photos(1)

Key Documents

804002

Sigma-Aldrich

Dow Corning® 736 heat resistant sealant

Synonym(s):

PDMS, Polydimethylsiloxane, Silicone Rubber, Silicone Sealant, Silicone elastomer

Sign Into View Organizational & Contract Pricing

Select a Size

3 EA
$136.00

$136.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
3 EA
$136.00

About This Item

UNSPSC Code:
12162002
NACRES:
NA.23

$136.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

form

paste

Quality Level

color

red

storage temp.

2-8°C

Application

736 Heat Resistant Sealant is a one-part nonslumping paste that cures to a rubbery solid at room temperature on exposure to water vapor in the air. This silicone product is formulated to perform at temperatures ranging from -65 to 260°C (-85 to 500°F) for continuous operation and to 315°C (600°F) for intermittent exposure.
The high temperature properties of this sealant make it ideally suited for:
  • Sealing and encapsulating heating elements in appliances.
  • Aerospace gasketing.
  • Moving oven belts.
  • Industrial ovens.
  • Bag filters on smoke stacks.
  • Other critical bonding, sealing, potting, encapsulating and protective coatings are parts which perform at high temperatures.

Packaging

3 ea of 10cc syringe applicator.

Legal Information

Dow Corning is a registered trademark of The Dow Chemical Company or an affiliated company of Dow

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service