Skip to Content
Merck
All Photos(1)

Key Documents

414026

Sigma-Aldrich

Trimethylsilyl polyphosphate

Synonym(s):

PPSE, Polyphosphoric acid trimethylsilyl ester

Sign Into View Organizational & Contract Pricing

Select a Size

25 ML
$167.00
100 ML
$515.00

$167.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 ML
$167.00
100 ML
$515.00

About This Item

Linear Formula:
{-P(=O)[OSi(CH3)3]O-}n
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

$167.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

form

liquid

Quality Level

refractive index

n20/D 1.434

density

1.18 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

C[Si](C)(C)OP(=O)=O

InChI

1S/C3H9O3PSi/c1-8(2,3)6-7(4)5/h1-3H3

InChI key

YSVQLWJDHYBITL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

  • Microwave-Assisted Synthesis of 2-Aryl-2-oxazolines, 5,6-Dihydro-4H-1,3-oxazines, and 4,5,6,7-Tetrahydro-1,3-oxazepines: Utilizes trimethylsilyl polyphosphate under solvent-free conditions for the efficient preparation of various oxazolines and oxazines (MC Mollo, LR Orelli, 2016).

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

39.2 °F - closed cup

Flash Point(C)

4 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tetrahedron Letters, 22, 1803-1803 (1981)
Tetrahedron Letters, 45, 2465-2465 (1989)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service