Skip to Content
Merck
All Photos(1)

Key Documents

481246

Sigma-Aldrich

Poly(dimethylsiloxane), bis(hydroxyalkyl) terminated

average Mn ~5,600

Synonym(s):

PDMS

Sign Into View Organizational & Contract Pricing

Select a Size

25 ML
$107.00
100 ML
$307.00

$107.00


Estimated to ship onMarch 29, 2025


Request a Bulk Order

Select a Size

Change View
25 ML
$107.00
100 ML
$307.00

About This Item

CAS Number:
MDL number:
UNSPSC Code:
12162002
NACRES:
NA.23

$107.00


Estimated to ship onMarch 29, 2025


Request a Bulk Order

mol wt

average Mn ~5,600

Quality Level

refractive index

n20/D 1.409 (lit.)

viscosity

100 cP(25 °C)(lit.)

hydroxyl value

20 mg KOH/g

density

0.98 g/mL at 25 °C (lit.)

Looking for similar products? Visit Product Comparison Guide

Application

Reactive resin modifier, paint additive and mold release agent.

Features and Benefits

Oxygen permeability.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Renxuan Xie et al.
Science advances, 6(46) (2020-11-15)
Super-soft elastomers derived from bottlebrush polymers show promise as advanced materials for biomimetic tissue and device applications, but current processing strategies are restricted to simple molding. Here, we introduce a design concept that enables the three-dimensional (3D) printing of super-soft

Articles

Self-assembled monolayers (SAMs) have diverse applications; article compares benefits of alkylthiolates on gold SAM systems.

Related Content

We offer PDMS surface modification kit which allows for durable hydrophilization of various polydimethylsiloxane (PDMS) surfaces.

Questions

  1. Dear Madam/Sir, Do you have PDI value for this compound Poly(dimethylsiloxane), bis(hydroxyalkyl) teminated by a GPC measurement? If possible, could we get some data relevant to elemental analysis, especially for the heavy metal analysis ?Thank! kg

    1 answer
    1. Unfortunately, this product does not have GPC measurement or elemental analysis data available. All the quality control tests are listed in the Certificate of Analysis.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service