Skip to Content
Merck
All Photos(1)

Key Documents

481955

Sigma-Aldrich

Poly(dimethylsiloxane), hydroxy terminated

viscosity ~65 cSt 

Synonym(s):

PDMS

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
$123.00
500 ML
$439.00

$123.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 ML
$123.00
500 ML
$439.00

About This Item

CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

$123.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

refractive index

n20/D 1.406

viscosity

~65 cSt

bp

>200 °C

density

0.97 g/mL at 25 °C

SMILES string

C[Si](C)(O)O

InChI

1S/C2H8O2Si/c1-5(2,3)4/h3-4H,1-2H3

InChI key

XCLIHDJZGPCUBT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

  • Effects of surface modification with hydroxyl terminated polydimethylsiloxane on the corrosion protection of polyurethane coating: This study investigates the improvement of corrosion resistance in polyurethane coatings through modification with hydroxy-terminated PDMS, highlighting its effectiveness in enhancing protective properties (JH Jeon, MY Shon, 2014).
  • Fabrication of silicone modified polyurethane matrix superhydrophobic coating with hydroxy-terminated polydimethylsiloxane modified SiO2 nanoparticles: The study explores the use of hydroxy-terminated PDMS in creating superhydrophobic surfaces for coatings, showing its utility in advanced material applications (Y Cui et al., 2022).
  • High glass transition temperature shape‐memory materials: Hydroxyl‐terminated polydimethylsiloxane‐modified cyanate ester: This research demonstrates the use of hydroxy-terminated PDMS to modify cyanate esters to create materials with high glass transition temperatures and shape-memory properties (Z Li et al., 2020).
  • The preparation and properties study of polydimethylsiloxane-based coatings modified by epoxy resin: This paper discusses the modification of PDMS with epoxy resin to enhance the performance of coatings, providing insights into its potential for creating more durable and effective surface treatments (C Zhou et al., 2016).

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Self-assembled monolayers (SAMs) have diverse applications; article compares benefits of alkylthiolates on gold SAM systems.

Related Content

We offer PDMS surface modification kit which allows for durable hydrophilization of various polydimethylsiloxane (PDMS) surfaces.

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service