Skip to Content
Merck
All Photos(2)

Key Documents

733385

Sigma-Aldrich

(11-Mercaptoundecyl)-N,N,N-trimethylammonium bromide

Synonym(s):

(11-Mercaptoundecyl)trimethylammonium bromide

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
$810.00

$810.00


Estimated to ship onMay 26, 2025


Request a Bulk Order

Select a Size

Change View
500 MG
$810.00

About This Item

Empirical Formula (Hill Notation):
C14H32BrNS
CAS Number:
Molecular Weight:
326.38
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.23

$810.00


Estimated to ship onMay 26, 2025


Request a Bulk Order

form

solid

mp

113-118 °C

storage temp.

−20°C

SMILES string

[Br-].C[N+](C)(C)CCCCCCCCCCCS

InChI

1S/C14H31NS.BrH/c1-15(2,3)13-11-9-7-5-4-6-8-10-12-14-16;/h4-14H2,1-3H3;1H

InChI key

SCOCXONJTURWMB-UHFFFAOYSA-N

General description

(11-Mercaptoundecyl)-N,N,N-trimethylammonium bromide (MABr) is an organothiol which forms a self-assembled monolayer (SAM) that facilitates the immobilization of a variety of nanoparticles.[1]

Application

MABr can be used to functionalize gold electrodes to fabricate sensing platform for bioelectronics-based applications.[2] It can also be used in surface modification of gold-platinum based nanocrystals (Au-Pt NCs) that find potential applications in biomedicine and bioengineering.[3]
This ammonium thiol molecule is reporated for the formation of photoconductive self-assembled monolayers.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Study of binary self-assembled monolayers of a novel anchoring thiol (11-mercaptoundecyl-N?, N ?, N?-trimethylammonium) in the electron transfer with glucose oxidase enzyme.
Baldo TA, et al.
Sensing and Bio-Sensing Research, 18(9), 37-44 (2018)
A halogen-free synthesis of gold nanoparticles using gold (III) oxide.
Sashuk V and Rogaczewski K
Journal of Nanoparticle Research, 18(9), 261-261 (2016)
Preventing UV induced cell damage by scavenging reactive oxygen species with enzyme-mimic Au-Pt nanocomposites.
Xiong B, et al.
Talanta, 120(9), 262-267 (2014)
Elzbieta Stepula et al.
ACS applied materials & interfaces, 12(29), 32321-32327 (2020-06-24)
There is an urgent clinical need for multicolor imaging of single cancer cells (no ensemble averaging) for identifying heterogenous expression of predictive biomarkers. Specifically, the comprehensive characterization of single disseminated tumor cells (sDTCs) responsible for metastatic relapse is the key
Thomas Keller et al.
Journal of biophotonics, 13(6), e201960126-e201960126 (2020-01-21)
The characteristic vibrational spectroscopic fingerprint of Raman reporter molecules adsorbed on noble metal nanoparticles is employed for the identification of target proteins by the corresponding surface-enhanced Raman scattering (SERS) nanotag-labeled antibodies. Here, we present the modular synthesis of thiolated polyenes

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service