Skip to Content
Merck
All Photos(1)

Key Documents

739294

Sigma-Aldrich

3-Amino-1-propanethiol hydrochloride

technical grade

Synonym(s):

(3-Mercaptopropyl)ammonium chloride

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
$474.00
500 MG
$785.00

$474.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025


Request a Bulk Order

Select a Size

Change View
250 MG
$474.00
500 MG
$785.00

About This Item

Linear Formula:
C3H10ClNS
CAS Number:
Molecular Weight:
127.64
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

$474.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025


Request a Bulk Order

grade

technical grade

form

solid

mp

80-85 °C

SMILES string

Cl.NCCCS

InChI

1S/C3H9NS.ClH/c4-2-1-3-5;/h5H,1-4H2;1H

InChI key

GMEDUXHKSSWXSL-UHFFFAOYSA-N

General description

3-Amino-1-propanethiol hydrochloride (APT) is an aminoalkylthiol that forms a self-assembled monolayer (SAM) of amine terminated alkanethiol. It can be used to form a spacer layer that provides a gap between the metal surface and nanoparticles.[1][2]

Application

APT can be used to form a plasmonic tunneling gap through coulomb blockage which can be potentially used in high-speed switching devices.[3][4][5] It can also form a SAM on gold nanorods(AuNR) for controlled spacing and photon absorption in photovoltaic and other photo-sensors based devices.[1]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Large scale solution assembly of quantum dot-gold nanorod architectures with plasmon enhanced fluorescence
Nepal D, et al.
ACS Nano, 7(10), 9064-9074 (2013)
Potential High-Speed Switching Nano-Device with Sub-Nanometer Gaps
2018 IEEE 13th Nanotechnology Materials & Devices Conference, 22(8), 1-4 (2018)
Switchable Metal-Insulator Phase Transition Metamaterials
Hajisalem G, et al.
Nano Letters, 17(5), 2940-2944 (2017)
Effect of surface roughness on self-assembled monolayer plasmonic ruler in nonlocal regime.
Hajisalem G, et al.
Optics Express, 22(8), 9604-9610 (2014)
Coulomb Blockade Plasmonic Switch.
Xiang D, et al.
Nano Letters, 17(4), 2584-2588 (2017)

Articles

Recent research highlights tunable properties of inorganic nanoparticles, driving interest in optoelectronics.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service