Skip to Content
Merck
All Photos(1)

Key Documents

674427

Sigma-Aldrich

11-Mercaptoundecanoic acid

98%

Synonym(s):

MUA, MUDA

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
$611.00

$611.00


Estimated to ship onMay 29, 2025


Request a Bulk Order

Select a Size

Change View
500 MG
$611.00

About This Item

Linear Formula:
HS(CH2)10CO2H
CAS Number:
Molecular Weight:
218.36
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

$611.00


Estimated to ship onMay 29, 2025


Request a Bulk Order

Assay

98%

form

solid

mp

46-50 °C (lit.)

storage temp.

2-8°C

SMILES string

OC(=O)CCCCCCCCCCS

InChI

1S/C11H22O2S/c12-11(13)9-7-5-3-1-2-4-6-8-10-14/h14H,1-10H2,(H,12,13)

InChI key

GWOLZNVIRIHJHB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

11-mercaptoundecanoic acid (MUA) has long alkane chains and carboxyl containing thiols that are highly packed and form a self-assembled monolayer (SAM). It modifies the surface of nanoparticles to facilitate their dispersion into water and solvent based mediums.[1][2]

Application

MUA is used for the carboxylic acid functionalization of 1-dodecanethiol-protected gold nanoparticles (AuNPs) by Murray place-displacement reaction generally utilized in biomedical applications.[3] It can also be used to modify the surface of platinum nanoparticles (PtNPs) by creating a ligand nano-assembly with a high surface coverage for use in surface-enhanced Raman spectroscopy (SERS).[4] A single cell biochip platform can be developed utilizing a cell attachment technique with MUA forming a SAM that was attached to single cells.[5] MUA may be used to prepare a tetrafluorophenyl-activated ester self-assembled monolayer to immobilize amine-modified oligonucleotides[6] and DNA.[7] It is also used to create 3D-assembled gold nanostructures for plasmonic biosensors.[8]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

No data available

Flash Point(C)

No data available

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yonghai Song et al.
Microscopy research and technique, 68(2), 59-64 (2005-10-18)
Immobilized DNA on preformed 11-mercaptoundecanoic acids (MUDA) self-assembled monolayers (SAMs) on a gold (111) surface was bound by a divalent cation bridges was imaged by atomic force microscopy (AFM). The DNA immobilization was attributed to the formation of ionic bridges
Thermogravimetry and evolved gas analysis for the investigation of ligand-exchange reaction in thiol-functionalized gold nanoparticles.
Locardi F, et al.
Journal of Analytical and Applied Pyrolysis, 132(5), 11-18 (2018)
Development of single cell biochip platform based on silicon for multiple patch clamping technology.
2009 IEEE 3rd International Conference on Nano/Molecular Medicine and Engineering, 29(6), 138-141 (2009)
Longhua Guo et al.
Analytical chemistry, 82(12), 5147-5153 (2010-05-18)
Three-dimensional gold nanoarchitecture was fabricated by layer-by-layer (LbL) deposition of gold nanoparticles (AuNPs) and multiwalled carbon nanotubes (MWCNTs) on a glass substrate for a highly sensitive plasmonic biosensor using a conventional UV-vis instrument. Carboxyl-functionalized MWCNTs were reacted with 3-mercaptopropyltriethoxysilane (MPTES)
Y Inoue et al.
Journal of the Royal Society, Interface, 5(25), 909-918 (2008-01-15)
A transfection array, which is specifically developed for use in high-throughput analyses of genome functions by the over-expression or suppression of genes on a chip, is expected to become an important method for post-genome research. High efficiency of gene expression

Articles

Recent research highlights tunable properties of inorganic nanoparticles, driving interest in optoelectronics.

Self-assembled monolayers (SAMs) have diverse applications; article compares benefits of alkylthiolates on gold SAM systems.

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service