Skip to Content
Merck
All Photos(1)

Key Documents

729159

Sigma-Aldrich

Poly(ethylene glycol) methyl ether thiol

average MN 6,000, chemical modification reagent thiol reactive, methoxy, thiol

Synonym(s):

Polyethylene glycol, Methoxy PEG thiol, Methoxypolyethylene glycol thiol, PEG thiol, mPEG thiol

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$394.00
5 G
$1,310.00

$394.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$394.00
5 G
$1,310.00

About This Item

Linear Formula:
CH3O(CH2CH2O)nCH2CH2SH
UNSPSC Code:
12162002
NACRES:
NA.23

$394.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Product Name

Poly(ethylene glycol) methyl ether thiol, average Mn 6,000

form

solid

mol wt

average Mn 6,000

reaction suitability

reagent type: chemical modification reagent
reactivity: thiol reactive

mp

56-61 °C

Mw/Mn

≤1.2

Ω-end

thiol

α-end

methoxy

polymer architecture

shape: linear
functionality: monofunctional

shipped in

dry ice

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

Poly(ethylene glycol) methyl ether thiol (mPEGthiol) is a thiol-terminated biocompatible polymer widely used in the surfacefunctionalization of gold nanoparticles. Thiol moieties bind with high affinityto gold surfaces by forming a self-assembled monolayer (SAM) through thecovalent thiolate–metal bond. mPEG thiol-coated Au nanoparticles findapplication in the fields of biosensors and drug delivery systems.

Application

Poly(ethylene glycol) methyl ether thiol-coated Au nanoparticles can be used for the determination of melamine T. by an optical method. It can also be used to fabricate switchable super-wetting membranes foroil/water separation.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Amina Ghorbel et al.
Bioelectrochemistry (Amsterdam, Netherlands), 137, 107642-107642 (2020-09-28)
The use of conductive nanoparticles (NPs) was previously proposed as a way to locally amplify the electric field (EF) intensity at the cell membrane to enhance cell electroporation. To achieve this, a close distance between the NPs and the cell
Jairo Pinto Oliveira et al.
Scientific reports, 9(1), 13859-13859 (2019-09-27)
Antibody-coated nanoparticles have recently attracted considerable attention, with the focus falling on diagnostics. Nevertheless, controlled antibody bioconjugation remains a challenge. Here, we present two strategies of bioconjugation with the aim of evaluating the best approach for the coupling of antibodies

Articles

Progress in biotechnology fields such as tissue engineering and drug delivery is accompanied by an increasing demand for diverse functional biomaterials. One class of biomaterials that has been the subject of intense research interest is hydrogels, because they closely mimic the natural environment of cells, both chemically and physically and therefore can be used as support to grow cells. This article specifically discusses poly(ethylene glycol) (PEG) hydrogels, which are good for biological applications because they do not generally elicit an immune response. PEGs offer a readily available, easy to modify polymer for widespread use in hydrogel fabrication, including 2D and 3D scaffold for tissue culture. The degradable linkages also enable a variety of applications for release of therapeutic agents.

Designing biomaterial scaffolds mimicking complex living tissue structures is crucial for tissue engineering and regenerative medicine advancements.

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service