Skip to Content
Merck
All Photos(1)

Key Documents

686220

Sigma-Aldrich

Hafnium carboxyethyl acrylate

60% in 1-butanol, contains 500 ppm methyl hydroquinone as inhibitor

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$687.00

$687.00


Estimated to ship onMay 26, 2025


Request a Bulk Order

Select a Size

Change View
25 G
$687.00

About This Item

Linear Formula:
Hf(CH2CHCO2(CH2)2CO2)4
Molecular Weight:
750.96
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

$687.00


Estimated to ship onMay 26, 2025


Request a Bulk Order

form

liquid

contains

500 ppm methyl hydroquinone as inhibitor

concentration

60% in 1-butanol

density

1.154 g/mL at 25 °C

SMILES string

[Hf+4].[O-]C(=O)CCOC(=O)C=C.[O-]C(=O)CCOC(=O)C=C.[O-]C(=O)CCOC(=O)C=C.[O-]C(=O)CCOC(=O)C=C

InChI

1S/4C6H8O4.Hf/c4*1-2-6(9)10-4-3-5(7)8;/h4*2H,1,3-4H2,(H,7,8);/q;;;;+4/p-4

InChI key

FGZZWYGTQYZLSX-UHFFFAOYSA-J

Application

Hafnium-containing multifunctional acrylate useful for producing cured, transparent films with high refractive indices.
Please see product no. 686204 for additional application and usage information (Aldrich Technical Bulletin AL-238 )

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system, Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

120.0 °F - closed cup

Flash Point(C)

48.89 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Polymeric materials, with unique refractive index properties, find applications in photonics and optical coatings for various devices.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service