Skip to Content
Merck
All Photos(2)

Key Documents

407232

Sigma-Aldrich

N-[(3-(Anilinomethylene)-2-chloro-1-cyclohexen-1-yl)methylene]aniline monohydrochloride

94%

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$977.00

$977.00


Estimated to ship onMay 20, 2025


Request a Bulk Order

Select a Size

Change View
5 G
$977.00

About This Item

Empirical Formula (Hill Notation):
C20H19ClN2 · HCl
CAS Number:
Molecular Weight:
359.29
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

$977.00


Estimated to ship onMay 20, 2025


Request a Bulk Order

Quality Level

Assay

94%

form

powder, crystals or chunks

mp

224 °C (dec.) (lit.)

εmax

>80000 at 524-530 nm in glacial acetic acid at 0.003 g/L

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

Cl[H].ClC1=C(CCC\C1=C/Nc2ccccc2)\C=N\c3ccccc3

InChI

1S/C20H19ClN2.ClH/c21-20-16(14-22-18-10-3-1-4-11-18)8-7-9-17(20)15-23-19-12-5-2-6-13-19;/h1-6,10-15,22H,7-9H2;1H/b16-14+,23-15+;

InChI key

VFKQWKQWRNVXTA-LPYKZFQHSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service