Skip to Content
Merck
All Photos(1)

Key Documents

407585

Sigma-Aldrich

2-Carboxyethyl acrylate oligomers

anhydrous, n=0-3, average MW~170

Synonym(s):

β-(Acryloyloxy)propionic acid, 2-Carboxyethyl acrylate, 3-(Acryloyloxy)propionic acid, Acrylic acid 2-carboxyethyl ester

Sign Into View Organizational & Contract Pricing

Select a Size

250 ML
$206.00

$206.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
250 ML
$206.00

About This Item

Linear Formula:
C3H3O2(C3H4O2)nH
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

$206.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

grade

anhydrous

form

liquid

contains

2000 ppm MEHQ as inhibitor

density

1.16 g/mL at 25 °C (lit.)

SMILES string

OC(=O)CCOC(=O)C=C

InChI

1S/C6H8O4/c1-2-6(9)10-4-3-5(7)8/h2H,1,3-4H2,(H,7,8)

InChI key

CYUZOYPRAQASLN-UHFFFAOYSA-N

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

212.0 °F - closed cup

Flash Point(C)

100 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The effect of acrylic acid dimer on the properties of carboxycement.
S Pokorný et al.
Journal of dentistry, 16(2), 76-79 (1988-04-01)
Mi-Sook Kim et al.
Journal of biomedical materials research. Part A, 83(3), 674-682 (2007-05-29)
We developed and characterized a novel in situ chitosan-poly(ethylene oxide) (PEO) hydrogel via two steps: 2-carboxyethyl acrylate molecules were grafted to the primary amine functional groups in chitosan in the first step and then Michael type addition reaction was processed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service