Skip to Content
Merck
All Photos(1)

Key Documents

408336

Sigma-Aldrich

Ethylene glycol phenyl ether acrylate

contains 75-125 ppm hydroquinone as inhibitor, 0-120 ppm hydroquinone monomethyl as inhibitor

Synonym(s):

2-Phenoxyethyl acrylate

Sign Into View Organizational & Contract Pricing

Select a Size

250 ML
$136.00
1 L
$424.00

$136.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
250 ML
$136.00
1 L
$424.00

About This Item

Linear Formula:
H2C=CHCO2CH2CH2OC6H5
CAS Number:
Molecular Weight:
192.21
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

$136.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

form

liquid

Quality Level

contains

0-120 ppm hydroquinone monomethyl as inhibitor
75-125 ppm hydroquinone as inhibitor

refractive index

n20/D 1.518 (lit.)

bp

84 °C/0.2 mmHg (lit.)

density

1.104 g/mL at 25 °C (lit.)

SMILES string

C=CC(=O)OCCOc1ccccc1

InChI

1S/C11H12O3/c1-2-11(12)14-9-8-13-10-6-4-3-5-7-10/h2-7H,1,8-9H2

InChI key

RZVINYQDSSQUKO-UHFFFAOYSA-N

General description

Ethylene glycol phenyl ether acrylate (PA) is a monofunctional acrylate.

Application

Ethylene glycol phenyl ether acrylate (PA) monomer may be used to fabricate polymerization induced grating pattern on photopolymer films. [1]

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fluorescent grating patterns of photopolymer film containing ethylene glycol phenyl ether acrylate.
Oh H, et al.
Journal of Nanoscience and Nanotechnology, 8(9), 4616-4620 (2008)

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service