Skip to Content
Merck
All Photos(1)

Key Documents

667676

Sigma-Aldrich

2-(2,4-Difluorophenyl)pyridine

97%

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$102.00

$102.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onMay 31, 2025


Request a Bulk Order

Select a Size

Change View
1 G
$102.00

About This Item

Empirical Formula (Hill Notation):
C11H7F2N
CAS Number:
Molecular Weight:
191.18
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$102.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onMay 31, 2025


Request a Bulk Order

Assay

97%

refractive index

n20/D 1.570

bp

95 °C/0.4 mmHg

density

1.254 g/mL at 25 °C

functional group

fluoro

SMILES string

Fc1ccc(c(F)c1)-c2ccccn2

InChI

1S/C11H7F2N/c12-8-4-5-9(10(13)7-8)11-3-1-2-6-14-11/h1-7H

InChI key

SSABEFIRGJISFH-UHFFFAOYSA-N

Application

Ligand used in a blue-light emitting Ir(III) complexes suitable for use as phosphorescent OLEDs.[1]

related product

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

217.9 °F

Flash Point(C)

103.3 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Blue-emitting heteroleptic iridium(III) complexes suitable for high-efficiency phosphorescent OLEDs.
Cheng-Han Yang et al.
Angewandte Chemie (International ed. in English), 46(14), 2418-2421 (2007-02-24)
Kin-Cheung Chan et al.
Dalton transactions (Cambridge, England : 2003), 44(34), 15135-15144 (2015-05-06)
A new series of cyclometalated iridium(III) complexes with isocyanoborate ligands [Ir(R2ppy)2(L)(CNBR'3)] (R = H or F; L = CNC6H4Cl-4 or PPh3; R' = Ph, C6F5 or C6H4Cl-4), [Ir(biqb)(ppy)(CNBR''3)] (R'' = C6F5 or C6H4Cl-4) and {Ir(ppy)2(CN)n[CNB(C6F5)3]2-n}(-) (n = 0 or 1)
Wendell Ho-Tin Law et al.
ChemMedChem, 9(6), 1316-1329 (2014-03-20)
We present a new class of phosphorescent cyclometalated iridium(III) bipyridyl-phenylenediamine complexes [Ir(N^C)2 (bpy-DA)](PF6 ) (bpy-DA=4-(N-(2-amino-5-methoxyphenyl)aminomethyl)-4'-methyl-2,2'-bipyridine; HN^C=2-(2,4-difluorophenyl)pyridine (Hdfppy) (1 a), 2-phenylpyridine (Hppy) (2 a), 2-phenylquinoline (Hpq) (3 a), 2-phenylcinchoninic acid methyl ester (Hpqe) (4 a)) and their triazole counterparts [Ir(N^C)2 (bpy-T)](PF6

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service