Skip to Content
Merck
All Photos(1)

Key Documents

658383

Sigma-Aldrich

Dichlorotetrakis(2-(2-pyridinyl)phenyl)diiridium(III)

Synonym(s):

[(ppy)2IrCl]2

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
$392.00

$392.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
500 MG
$392.00

About This Item

Empirical Formula (Hill Notation):
C44H32Cl2Ir2N4
CAS Number:
Molecular Weight:
1072.09
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

$392.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

form

solid

Quality Level

reaction suitability

core: iridium
reaction type: Photocatalysis
reagent type: catalyst

photocatalyst activation

465 nm

SMILES string

Cl[Ir](c1ccccc1-c2ccccn2)c3ccccc3-c4ccccn4.Cl[Ir](c5ccccc5-c6ccccn6)c7ccccc7-c8ccccn8

InChI

1S/4C11H9N.2ClH.2Ir/c4*1-2-6-10(7-3-1)11-8-4-5-9-12-11;;;;/h4*1-9H;2*1H;;/q;;;;;;2*+1/p-2

InChI key

NARZJSRZFGZBIZ-UHFFFAOYSA-L

Application

Precursor for several OLED Iridium compounds. Has some use as a catalyst and in OLEDs.[1]

Product can be used with our line of photoreactors: Including Penn PhD (Z744035) & SynLED 2.0 (Z744080)

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Journal of the American Chemical Society, 106, 6647-6647 (1984)

Articles

Since their discovery, organic light emitting devices (OLEDs) have evolved from a scientific curiosity into a technology with applications in flat panel displays and the potential to revolutionize the lighting market. During their relatively short history, the technology has rapidly advanced, and device efficiencies have increased more than 20-fold, approaching the theoretical limit for internal quantum efficiencies.

Related Content

Explore reliable, premium grade catalysis materials for your pharma or industrial project. Specialty chemicals and formulations are available in bulk quantities and volumes from a few grams to multi-metric tons with complete documentation to simplify your leap from development to commercialization.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service