Skip to Content
Merck
All Photos(1)

Key Documents

751464

Sigma-Aldrich

Bis[2-(4,6-difluorophenyl)pyridinato-C2,N](picolinato)iridium(III)

97%

Synonym(s):

F2Irpic, FIrpic

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
$275.00

$275.00


Estimated to ship onApril 16, 2025


Request a Bulk Order

Select a Size

Change View
250 MG
$275.00

About This Item

Empirical Formula (Hill Notation):
C28H16F4IrN3O2
CAS Number:
Molecular Weight:
694.66
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

$275.00


Estimated to ship onApril 16, 2025


Request a Bulk Order

Quality Level

Assay

97%

form

solid

mp

330-335 °C

Orbital energy

HOMO 5.8 eV 
LUMO 2.9 eV 

OLED Device Performance

ITO/NPD/mCP/BCPO:FIrpic (8%)/TAZ/LiF/Al

  • Color: blue
  • Max. Luminance: 35677 Cd/m2
  • Max. EQE: 23.5 %
  • Turn-On Voltage: 2.8 V

ITO/PEDOT:PSS/BCC-27: FIrpic (10 wt%)/TAZ/Cs2CO3:Al
  • Color: blue
  • Max. Luminance: 19200 Cd/m2
  • Max. EQE: 9.7 %
  • Turn-On Voltage: 3.9 V

ITO/PEDOT:PSS/NPD/TCTA/m-CBTZ:FIrpic (10 wt%)/DPPS/TAZ/LiF/Al
  • Color: blue
  • Max. Luminance: 29500 Cd/m2
  • Max. EQE: 8.8 %
  • Turn-On Voltage: 3.5 V

ITO/PEDOT:PSS/NPD/TCTA/p-CBTZ:FIrpic (10 wt%)/DPPS/TAZ/LiF/Al
  • Color: blue
  • Max. Luminance: 15500 Cd/m2
  • Max. EQE: 8 %
  • Turn-On Voltage: 3.5 V

SMILES string

Fc1cc(F)c(-c2ccccn2)c(c1)[Ir](OC(=O)c3ccccn3)c4cc(F)cc(F)c4-c5ccccn5

InChI

1S/2C11H6F2N.C6H5NO2.Ir/c2*12-8-4-5-9(10(13)7-8)11-3-1-2-6-14-11;8-6(9)5-3-1-2-4-7-5;/h2*1-4,6-7H;1-4H,(H,8,9);/q;;;+1/p-1

InChI key

CTOBHCLNNFAQNN-UHFFFAOYSA-M

Application

FIrpic is a light blue heteroleptic triplet emitter.This material is typically doped into host matrices such as tetra-aryl silanes and short conjugation length carbazole derivatives in OLEDs; showing a good quantum efficiency.[1]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Suppression of efficiency roll off in blue phosphorescent organic light-emitting devices using double emission layers with additional carrier-transporting material
Lee, M.; Lin, J.; Chu, M.; et al.
Applied Physics Letters, 94, 083506-083506 (2009)
Fluorine cleavage of the light blue heteroleptic triplet emitter FIrpic
Sivasubramaniam V, et al.
Journal of Fluorine Chemistry, 130(7), 640-649 (2009)
Cai, X.Y.; Padmaperuma, A.B.; Sapochak, L.S.; Vecchi, P.A.; Burrows, P.E.
Applied Physics Letters, 92, 083308-083308 (2008)
High-triplet-energy tri-carbazole derivatives as host materials for efficient solution-processed blue phosphorescent devices
Jiang, W.; et al.
Journal of Materials Chemistry, 21, 4918 - 4926 (2011)
Phenylcarbazole-dipyridyl triazole hybrid as bipolar host material for phosphorescent OLEDs
Hung, W.; et al.
Journal of Materials Chemistry, 22, 5410-5418 (2012)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service