Skip to Content
Merck
All Photos(2)

Documents

198870

Sigma-Aldrich

2-(p-Tolyl)pyridine

≥97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H11N
CAS Number:
Molecular Weight:
169.22
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥97%

form

liquid

refractive index

n20/D 1.617 (lit.)

bp

170-180 °C/20 mmHg (lit.)

density

0.99 g/mL at 25 °C (lit.)

SMILES string

Cc1ccc(cc1)-c2ccccn2

InChI

1S/C12H11N/c1-10-5-7-11(8-6-10)12-4-2-3-9-13-12/h2-9H,1H3

InChI key

KJNZQKYSNAQLEO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

The chloro complexes of nickel, copper and cobalt with 2-(p-tolyl)pyridine have been prepared.

Application

2-(p-Tolyl)pyridine may be used as a pharmaceutical intermediate.

related product

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E G Vlakh et al.
Scientific reports, 7, 41991-41991 (2017-02-06)
The growing attention to the luminescent nanocarriers is strongly stimulated by their potential application as drug delivery systems and by the necessity to monitor their distribution in cells and tissues. In this communication we report on the synthesis of amphiphilic
Thermal, spectral and magnetic studies of some first row transition metal complexes of 2-(p-tolyl) pyridine.
Allan JR, et al.
Thermochimica Acta, 160(2), 329-335 (1990)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service