Skip to Content
Merck
All Photos(1)

Key Documents

637408

Sigma-Aldrich

Methyl 4-vinylbenzoate

97%

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$211.00
5 G
$679.00

$211.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$211.00
5 G
$679.00

About This Item

Empirical Formula (Hill Notation):
C10H10O2
CAS Number:
Molecular Weight:
162.19
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$211.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

97%

form

solid

mp

32-37 °C (lit.)

functional group

ester

storage temp.

2-8°C

SMILES string

COC(=O)c1ccc(C=C)cc1

InChI

1S/C10H10O2/c1-3-8-4-6-9(7-5-8)10(11)12-2/h3-7H,1H2,2H3

InChI key

NUMHUJZXKZKUBN-UHFFFAOYSA-N

Related Categories

Application

Methyl 4-vinylbenzoate can be used in the synthesis of vinyl monomers of bis-crowned malachite green leuconitrile, which can find applications in photoresponsive ionic conduction.[1] The co-polymer formed by reacting methyl 4-vinylbenzoate and (4-trimethylsilyl)styrene can be oxidized to form polyradicals with good membrane forming ability.[2]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

"Synthesis of galvinoxyl unit-containing derivatives of poly (phenylacetylene) and polystyrene, and oxygen permeation behavior of their membranes"
Kaneko.T, et al.
Polymer, 41(12), 4437-4444 (2000)
"Cation complexation and photochromism of copolymers carrying pendant crowned malachite green moiety"
Kimura K, et al.
Macromolecules, 34(07), 2262-2268 (2001)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service