Skip to Content
Merck
All Photos(1)

Key Documents

527904

Sigma-Aldrich

Vinyl cinnamate

≥95%

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
$253.00

$253.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 ML
$253.00

About This Item

Linear Formula:
C6H5CH=CHCO2CH=CH2
CAS Number:
Molecular Weight:
174.20
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

$253.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

≥95%

contains

50 ppm MEHQ as inhibitor

storage temp.

2-8°C

SMILES string

C=COC(=O)\C=C\c1ccccc1

InChI

1S/C11H10O2/c1-2-13-11(12)9-8-10-6-4-3-5-7-10/h2-9H,1H2/b9-8+

InChI key

WGXGKXTZIQFQFO-CMDGGOBGSA-N

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Rongling Yang et al.
Frontiers in bioengineering and biotechnology, 8, 273-273 (2020-05-05)
Highly efficient and regioselective synthesis of pharmacologically interesting aromatic esters of arbutin catalyzed by immobilized lipase from Penicillium expansum in co-solvent systems was successfully carried out. As compared to tetrahydrofuran solvent, the initial rate and substrate conversion of arbutin vanilylation
Jérémie Brand et al.
Carbohydrate polymers, 169, 189-197 (2017-05-16)
Herein we propose a versatile method for the surface tailoring of cellulose nanocrystals (CNCs) based on the reactivity of vinyl ester molecules toward the accessible hydroxyl groups located at the surface of the nanoparticles. CNCs produced from wood pulp were

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service