Skip to Content
Merck
All Photos(1)

Documents

455644

Sigma-Aldrich

4-tert-Butoxystyrene

99%, contains 200 ppm 4-tert-butylcatechol as inhibitor

Synonym(s):

4-tert -Butoxystyrene, p -tert -Butoxystyrene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2C=CHC6H4OC(CH3)3
CAS Number:
Molecular Weight:
176.25
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Assay

99%

contains

200 ppm 4-tert-butylcatechol as inhibitor

refractive index

n20/D 1.524 (lit.)

bp

72-73 °C/0.1 mmHg (lit.)

mp

−38 °C (lit.)

density

0.936 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

CC(C)(C)Oc1ccc(C=C)cc1

InChI

1S/C12H16O/c1-5-10-6-8-11(9-7-10)13-12(2,3)4/h5-9H,1H2,2-4H3

InChI key

GRFNSWBVXHLTCI-UHFFFAOYSA-N

Related Categories

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

206.6 °F - closed cup

Flash Point(C)

97 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wei-Cheng Chen et al.
Polymers, 12(2) (2020-02-23)
In this study, we synthesized two main chain-type block copolymers featuring hydrogen bond donor and acceptor segments through atom transfer radical polymerization (ATRP) using a bifunctionalized polyhedral oligomeric silsesquioxane (POSS) nanoparticle as the initiator. Hydrosilylation of vinylbenzyl chloride at the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service