Skip to Content
Merck
All Photos(1)

Key Documents

380547

Sigma-Aldrich

4-Acetoxystyrene

96%, contains 200-300 ppm monomethyl ether hydroquinone as inhibitor

Synonym(s):

4-Ethenylphenol acetate, 4-Vinylphenyl acetate

Sign Into View Organizational & Contract Pricing

Select a Size

25 ML
$184.00

$184.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025


Request a Bulk Order

Select a Size

Change View
25 ML
$184.00

About This Item

Linear Formula:
CH3CO2C6H4CH=CH2
CAS Number:
Molecular Weight:
162.19
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

$184.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025


Request a Bulk Order

Assay

96%

form

liquid

contains

200-300 ppm monomethyl ether hydroquinone as inhibitor

refractive index

n20/D 1.538 (lit.)

viscosity

4.2 cP(25 °C, Brookfield)(lit.)

bp

260 °C (lit.)

mp

7-8 °C (lit.)

density

1.06 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

CC(=O)Oc1ccc(C=C)cc1

InChI

1S/C10H10O2/c1-3-9-4-6-10(7-5-9)12-8(2)11/h3-7H,1H2,2H3

InChI key

JAMNSIXSLVPNLC-UHFFFAOYSA-N

Specificity

Reactivity ratio: 4-Acetoxystyrene/styrene 1.02/0.80
4-Acetoxystyrene/methyl methacrylate 0.76/0.88
4-Acetoxystyrene/maleic anhydride 0/0

Application

Polymer used in microlithography. Precursor to p-hydroxystyrene that is readily derivatized.

Features and Benefits

Undergoes free radical polymerization, similar to styrene

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

190.4 °F - closed cup

Flash Point(C)

88 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Articles

Block copolymer synthesis using a commercially available nitroxide-mediated radical polymerization (NMP) initiator

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service