Skip to Content
Merck
All Photos(1)

Key Documents

510297

Sigma-Aldrich

5-Bromo-2-methoxypyridine

95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H6BrNO
CAS Number:
Molecular Weight:
188.02
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Assay:
95%

Assay

95%

refractive index

n20/D 1.555 (lit.)

bp

80 °C/12 mmHg (lit.)

density

1.453 g/mL at 25 °C (lit.)

functional group

bromo

SMILES string

COc1ccc(Br)cn1

InChI

1S/C6H6BrNO/c1-9-6-3-2-5(7)4-8-6/h2-4H,1H3

InChI key

XADICJHFELMBGX-UHFFFAOYSA-N

Application

A building block for the β-alanine moiety of an αvβ3 antagonist[1][2] and for the synthesis of a potent and selective somatostatin sst3 receptor antagonist.[3]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

204.8 °F - closed cup

Flash Point(C)

96 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Nobuyoshi Yasuda et al.
The Journal of organic chemistry, 69(6), 1959-1966 (2004-04-03)
A practical preparation of an alpha(v)beta(3) antagonist is reported. The antagonist consists of three key components, a tetrahydronaphthyridine moiety, a beta-alanine moiety, and a central imidazolidone moiety. The tetrahydronaphthyridine component was prepared using two different methods, both of which relied
A scaleable synthesis of methyl 3-amino-5-(4-fluorobenzyl)-2-pyridinecarboxylate.
Organic Process Research & Development 11.5 (2007): 899-902
Boros EE, et al.
Organic Process Research & Development, 11(5), 899-902 (2007)
Negishi Cross-Coupling Reactions Catalyzed by an Aminophosphine-Based Nickel System: A Reliable and General Applicable Reaction Protocol for the High-Yielding Synthesis of Biaryls.
Gerber R and Frech CM.
Chemistry (Weinheim An Der Bergstrasse, Germany), 17(42), 11893-11904 (2011)
Tetrahedron Asymmetry, 14, 3469-3469 (2003)
Synthesis and biological evaluation of 1-(benzenesulfonamido)-2-[5-(N-hydroxypyridin-2 (1H)-one)] acetylene regioisomers: A novel class of 5-lipoxygenase inhibitors.
Chowdhury MA, et al.
Bioorganic & Medicinal Chemistry Letters, 18(14), 4195-4198 (2008)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service