Skip to Content
Merck
All Photos(1)

Key Documents

674761

Sigma-Aldrich

(R)-1-Boc-3-methylpiperazine

97%

Synonym(s):

tert-Butyl (R)-3-methyl-1-piperazinecarboxylate

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$118.00

$118.00


Estimated to ship onMarch 21, 2025


Request a Bulk Order

Select a Size

Change View
1 G
$118.00

About This Item

Empirical Formula (Hill Notation):
C10H20N2O2
CAS Number:
Molecular Weight:
200.28
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

$118.00


Estimated to ship onMarch 21, 2025


Request a Bulk Order

Quality Level

Assay

97%

storage temp.

-10 to -25°C

SMILES string

C[C@@H]1CN(CCN1)C(=O)OC(C)(C)C

InChI

1S/C10H20N2O2/c1-8-7-12(6-5-11-8)9(13)14-10(2,3)4/h8,11H,5-7H2,1-4H3/t8-/m1/s1

InChI key

FMLPQHJYUZTHQS-MRVPVSSYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

223.0 °F

Flash Point(C)

106.1 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service