Skip to Content
Merck
All Photos(1)

Key Documents

520535

Sigma-Aldrich

2-Bromo-6-methoxypyridine

97%

Sign Into View Organizational & Contract Pricing

Select a Size

5 ML
$119.00

$119.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 ML
$119.00

About This Item

Empirical Formula (Hill Notation):
C6H6BrNO
CAS Number:
Molecular Weight:
188.02
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$119.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

97%

refractive index

n20/D 1.559 (lit.)

bp

206 °C (lit.)

density

1.53 g/mL at 25 °C (lit.)

functional group

bromo

SMILES string

COc1cccc(Br)n1

InChI

1S/C6H6BrNO/c1-9-6-4-2-3-5(7)8-6/h2-4H,1H3

InChI key

KMODISUYWZPVGV-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

219.9 °F - closed cup

Flash Point(C)

104.4 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of 2, 2'-Bipyridines: Versatile Building Blocks for Sexy Architectures and Functional Nanomaterials.
Newkome GR, et al.
European Journal of Organic Chemistry, 2, 235-254 (2004)
Robert T. Jonas et al.
Inorganic chemistry, 37(26), 6615-6629 (2001-10-24)
An efficient modular protocol for synthesizing a series of facial-capping tris-pyridyl ligands, based on the tris(2-pyridyl)methoxymethane backbone, has been developed which allows for systematic variations of the steric demands at the periphery of the ligand. The coordination chemistry of one
A convenient synthesis of cyclopenta [b] pyridin-2, 5-dione as a non-glycosidic cardiotonic agent.
Robert N, et al.
ARKIVOC (Gainesville, FL, United States), 7, 92-100 (2008)
Mohammed K Elmkaddem et al.
Chemical communications (Cambridge, England), 46(6), 925-927 (2010-01-29)
A copper(i) catalyzed amination reaction utilizing aqueous ammonia and operating under mild conditions is presented. This method was employed for the efficient synthesis of various aminopyridine derivatives bearing electron withdrawing and electron donating groups.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service