Skip to Content
Merck
All Photos(3)

Key Documents

683345

Sigma-Aldrich

[Pd2(dba)3] x dba

Umicore

Synonym(s):

Tris(dibenzylideneacetone)dipalladium(0), Pd2dba3, Pd2(dba)3

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
$68.00
5 G
$506.00
50 G
$2,530.00

$68.00


Estimated to ship onMarch 13, 2025


Request a Bulk Order

Select a Size

Change View
500 MG
$68.00
5 G
$506.00
50 G
$2,530.00

About This Item

Linear Formula:
(C6H5CH=CHCOCH=CHC6H5)3Pd2
CAS Number:
Molecular Weight:
915.72
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

$68.00


Estimated to ship onMarch 13, 2025


Request a Bulk Order

Quality Level

form

powder

reaction suitability

core: palladium
reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Cross Couplings
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: catalyst

mp

152-155 °C (lit.)

storage temp.

2-8°C

SMILES string

[Pd].[Pd].O=C(\C=C\c1ccccc1)/C=C/c2ccccc2.O=C(\C=C\c3ccccc3)/C=C/c4ccccc4.O=C(\C=C\c5ccccc5)/C=C/c6ccccc6

InChI

1S/3C17H14O.2Pd/c3*18-17(13-11-15-7-3-1-4-8-15)14-12-16-9-5-2-6-10-16;;/h3*1-14H;;/b3*13-11+,14-12+;;

InChI key

CYPYTURSJDMMMP-WVCUSYJESA-N

Looking for similar products? Visit Product Comparison Guide

Application

Umicore Precatalysts for Asymmetric and Cross-Coupling Catalysis

Catalyst for C-C and C-N bond formation especially Suzuki, Heck coupling reaction

Legal Information

Product of Umicore

Additional information available at www.pmc.umicore.com

related product

Product No.
Description
Pricing

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

George R Kracke et al.
ChemMedChem, 10(1), 62-67 (2014-11-26)
Clinically there is a need for local anesthetics with a greater specificity of action on target cells and longer duration. We have synthesized a series of local anesthetic derivatives we call boronicaines in which the aromatic phenyl ring of lidocaine
Michael J Ardolino et al.
Tetrahedron, 71(37), 6409-6413 (2015-08-19)
While Pd-catalyzed allyl-allyl cross-couplings in the presence of small-bite-angle bidentate ligands reliably furnish the branched regioisomer with high levels of selectivity, cross-couplings in the presence of large-bite-angle bidentate ligands give varying, often unpredictable, levels of selectivity. In a combined computational
Zhouen Zhang et al.
Journal of labelled compounds & radiopharmaceuticals, 57(8), 540-549 (2014-07-06)
The nucleosides zidovudine (AZT), stavudine (d4T), and telbivudine (LdT) are approved for use in the treatment of human immunodeficiency virus (HIV) and hepatitis B virus (HBV) infections. To promote positron emission tomography (PET) imaging studies on their pharmacokinetics, pharmacodynamics, and

Articles

Precatalysts for asymmetric catalysis offer batch-to-batch consistency in a range of catalytic reactions, supporting diverse research needs.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service