Skip to Content
Merck
All Photos(1)

Key Documents

491853

Sigma-Aldrich

Benzo[b]fluoranthene-d12

98 atom % D

Synonym(s):

Benz[e]acephenanthrylene-d12

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$1,030.00

$1,030.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request more information

Select a Size

Change View
10 MG
$1,030.00

About This Item

Empirical Formula (Hill Notation):
C20D12
CAS Number:
Molecular Weight:
264.38
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.12

$1,030.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request more information

isotopic purity

98 atom % D

Quality Level

Assay

99% (CP)

form

solid

mp

163-165 °C (lit.)

mass shift

M+12

SMILES string

[2H]c1c([2H])c([2H])c-2c(c1[2H])-c3c([2H])c([2H])c([2H])c4c5c([2H])c([2H])c([2H])c([2H])c5c([2H])c-2c34

InChI

1S/C20H12/c1-2-7-14-13(6-1)12-19-16-9-4-3-8-15(16)18-11-5-10-17(14)20(18)19/h1-12H/i1D,2D,3D,4D,5D,6D,7D,8D,9D,10D,11D,12D

InChI key

FTOVXSOBNPWTSH-AQZSQYOVSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Health hazardEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Ana L L Dauner et al.
Journal of environmental management, 223, 417-425 (2018-06-27)
The concentration, distribution, and ecological risk of hydrocarbons, as well as bulk parameters, were determined in surface sediments of the Babitonga Bay, a subtropical human-impacted estuary in South Atlantic. Total aliphatic and polycyclic aromatic hydrocarbons (PAHs) ranged between 0.8 and
Weier Hao et al.
Food additives & contaminants. Part A, Chemistry, analysis, control, exposure & risk assessment, 37(7), 1202-1215 (2020-05-05)
In this work, we describe a method developed to quantify persistent organic pollutants (POPs) including polycyclic aromatic hydrocarbons (PAHs) and organochlorine pesticides (OCPs) in dietary supplement samples using stir-bar sorptive extraction (SBSE)-GC-MS/MS-isotope dilution mass spectrometry (IDMS). This method enables accurate
Maria João Fraqueza et al.
Foods (Basel, Switzerland), 9(1) (2020-01-19)
The manufacturing of dry-cured meat products usually includes a smoking step. Polycyclic aromatic hydrocarbons (PAHs) are potentially carcinogenic chemical compounds that may result from smoking. The aim of the present study was to optimize the smoking regime of traditional dry-cured

Articles

Compare PAH determination in olive oil: traditional GB 5009.265-2021 method vs. faster, simpler SPE with Supelclean EZ-POP NP; both meet criteria.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service