Skip to Content
Merck
All Photos(1)

Key Documents

451797

Sigma-Aldrich

Benzo[a]pyrene-d12

98 atom % D

Synonym(s):

Deuterated benzo[a]pyrene, 3,4-Benzopyrene-d12

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$388.00

$388.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request more information

Select a Size

Change View
10 MG
$388.00

About This Item

Empirical Formula (Hill Notation):
C20D12
CAS Number:
Molecular Weight:
264.38
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.12

$388.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request more information

isotopic purity

98 atom % D

Quality Level

Assay

99% (CP)

form

solid

bp

495 °C (lit.)

mp

177-180 °C (lit.)

application(s)

electronics

mass shift

M+12

storage temp.

room temp

SMILES string

[2H]c1c([2H])c([2H])c2c(c1[2H])c([2H])c3c([2H])c([2H])c4c([2H])c([2H])c([2H])c5c([2H])c([2H])c2c3c45

InChI

1S/C20H12/c1-2-7-17-15(4-1)12-16-9-8-13-5-3-6-14-10-11-18(17)20(16)19(13)14/h1-12H/i1D,2D,3D,4D,5D,6D,7D,8D,9D,10D,11D,12D

InChI key

FMMWHPNWAFZXNH-AQZSQYOVSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Benzo[a]pyrene-d12 is an isotope-labeled analog of the polycyclic aromatic hydrocarbon benzo[a]pyrene, wherein all the carbon attached protons (C-H) are replaced by deuterium atoms.

Application

Benzo[a]pyrene-d12 can be used as a stable isotope internal standard for the quantification of benzo(a)pyrene content in specific food samples using gas chromatography/mass spectrometry.[1]

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Signal Word

Danger

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B - Muta. 1B - Repr. 1B - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Spanish smoked meat products: benzo (a) pyrene (BaP) contamination and moisture
Ledesma E, et al.
Journal of food composition and analysis, 37, 87-94 (2015)
Inyang O Oyo-Ita et al.
Archives of environmental contamination and toxicology, 70(2), 372-382 (2015-11-08)
The distribution of aliphatic and aromatic hydrocarbons in surface sediments of the lower course of the Imo River (Nigeria) was investigated to determine the sources and fate of these compounds. The aliphatic fraction is characterized by a widespread contribution of
Wan-Ting Chang et al.
Marine pollution bulletin, 97(1-2), 217-223 (2015-06-16)
This work investigates the dependence of the sampling rate (Rs) of semi-permeable membrane devices (SPMDs) on flow rate and temperature. The in situ Rs values were obtained using performance reference compounds (PRCs) with weighted polynomial regression and used to estimate
Noor A Aly et al.
Journal of exposure science & environmental epidemiology, 31(5), 810-822 (2021-04-26)
Hurricane Florence made landfall in North Carolina in September 2018 causing extensive flooding. Several potential point sources of hazardous substances and Superfund sites sustained water damage and contaminants may have been released into the environment. This study conducted temporal analysis
Badreddine Barhoumi et al.
Marine environmental research, 101, 184-195 (2014-08-12)
In this study, biological responses and contaminant levels in biological tissues were investigated in grass goby fish specimens (Zosterisessor ophiocephalus) collected from five stations in a moderately polluted ecosystem, namely the Bizerte lagoon on the north coast of Tunisia. The

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service