Merck
  • Dry-Cured Meat Products According to the Smoking Regime: Process Optimization to Control Polycyclic Aromatic Hydrocarbons.
Dry-Cured Meat Products According to the Smoking Regime: Process Optimization to Control Polycyclic Aromatic Hydrocarbons.
Foods (Basel, Switzerland) (2020-01-19)
Maria João Fraqueza, Marta Laranjo, Susana Alves, Maria Helena Fernandes, Ana Cristina Agulheiro-Santos, Maria José Fernandes, Maria Eduarda Potes, Miguel Elias
ABSTRACT
The manufacturing of dry-cured meat products usually includes a smoking step. Polycyclic aromatic hydrocarbons (PAHs) are potentially carcinogenic chemical compounds that may result from smoking. The aim of the present study was to optimize the smoking regime of traditional dry-cured meat products in order to minimize the presence of PAHs. Dry-cured sausages were submitted to different smoking regimes: (A) no smoking; (B) 20 h effective smoking; (C) 60 h effective smoking; (D) effective smoking until reaching 38%-40% weight losses. Three independent batches were produced per smoking regime, and three samples per batch were analyzed. Microbiological, physicochemical, and sensory analyses were performed. The total PAHs content was generally low and did not differ significantly in meat products submitted to the four different smoking regimes. The PAH4 and benzo(α)pyrene levels were below the established legal limits in all analyzed dry-cured sausages. Nevertheless, non-smoked sausages always showed lower PAHs values for all PAHs groups.
Sigma-Aldrich
Acenaphthylene-d8, ≥98 atom % D, 98% (CP)
Pricing and availability is not currently available.
Sigma-Aldrich
Benzo[b]fluoranthene-d12, 98 atom % D
Pricing and availability is not currently available.