Skip to Content
Merck
All Photos(2)

Key Documents

482099

Sigma-Aldrich

Cyclopropanemethanol

≥99.5%

Synonym(s):

(Hydroxymethyl)cyclopropane, CPMO, Cyclopropyl carbinol, Cyclopropylmethanol

Sign Into View Organizational & Contract Pricing

Select a Size

25 ML
$146.00
250 ML
$385.00
1 L
$829.00

$146.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onMay 31, 2025


Request a Bulk Order

Select a Size

Change View
25 ML
$146.00
250 ML
$385.00
1 L
$829.00

About This Item

Linear Formula:
C3H5CH2OH
CAS Number:
Molecular Weight:
72.11
Beilstein:
1846846
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$146.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onMay 31, 2025


Request a Bulk Order

Assay

≥99.5%

refractive index

n20/D 1.431 (lit.)

bp

123-124 °C/738 mmHg (lit.)

density

0.89 g/mL at 25 °C (lit.)

functional group

hydroxyl

SMILES string

OCC1CC1

InChI

1S/C4H8O/c5-3-4-1-2-4/h4-5H,1-3H2

InChI key

GUDMZGLFZNLYEY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Cyclopropanemethanol (Cyclopropyl carbinol, CPMO), a cycloalkanemethanol, is an anaesthetic.[1] The coupling reaction of cyclopropanemethanol with alkynes to form substituted allylic alcohols has been reported.[2] The microwave spectrum of CPMO has been recorded. Its rotational constants and dipole moment have been determined.[3]

Application

Cyclopropanemethanol may be used in the preparation of:
  • cyclopropanecarbaldehyde[4]
  • cyclopropylmethylsulfonate[5]
  • dibenzyl cyclopropylmethyl phosphate[6]

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

95.0 °F - closed cup

Flash Point(C)

35 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The microwave spectrum, dipole moment, and structure of cyclopropyl carbinol.
Bhaumik A, et al.
Canadian Journal of Chemistry, 48(19) , 2949-2954 (1970)
Surbhi Soni et al.
Chirality, 30(1), 85-94 (2017-10-25)
A profoundly time-efficient chemoenzymatic method for the synthesis of (S)-3-(4-chlorophenoxy)propan-1,2-diol and (S)-1-chloro-3-(2,5-dichlorophenoxy)propan-2-ol, two important pharmaceutical intermediates, was successfully developed using Pseudomonas fluorescens lipase (PFL). Kinetic resolution was successfully achieved using vinyl acetate as acylating agent, toluene/hexane as solvent, and reaction
D E Raines et al.
Anesthesiology, 78(5), 918-927 (1993-05-01)
N-alkanols containing up to 12 carbons are anesthetic; however, those with more than 12 carbons are not. This phenomenon has been termed cutoff. Lipid disordering theories of anesthesia suggest that cutoff occurs because the alkyl chains of long-chain alcohols approach
Galvanic Deposition of Au on Paperlike Cu Fiber for High-Efficiency, Low-Temperature Gas-Phase Oxidation of Alcohols.
Zhao G, et al.
ChemCatChem, 3(10), 1629-1636 (2011)
Nickel-Catalyzed Redox-Economical Coupling of Alcohols and Alkynes to Form Allylic Alcohols.
Nakai K, et al.
Journal of the American Chemical Society, 136(22), 7797-7800 (2014)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service