Skip to Content
Merck
All Photos(2)

Key Documents

188204

Sigma-Aldrich

p-Toluenesulfonylmethyl isocyanide

98%

Synonym(s):

(p-Tolylsulfonyl)methyl isocyanide, TosMIC, Tosylmethyl isocyanide

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$51.60
25 G
$175.00

$51.60

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$51.60
25 G
$175.00

About This Item

Linear Formula:
CH3C6H4SO2CH2NC
CAS Number:
Molecular Weight:
195.24
Beilstein:
3592382
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$51.60

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

98%

form

solid

mp

109-113 °C (lit.)

solubility

water: slightly soluble

functional group

amine
isonitrile
sulfone

storage temp.

2-8°C

SMILES string

Cc1ccc(cc1)S(=O)(=O)C[N+]#[C-]

InChI

1S/C9H9NO2S/c1-8-3-5-9(6-4-8)13(11,12)7-10-2/h3-6H,7H2,1H3

InChI key

CFOAUYCPAUGDFF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

p-Toluenesulfonylmethyl isocyanide undergoes base-promoted 1,3-dipolar cycloaddition reaction with immobilized imines under microwave irradiation to yield 1,5-disubstituted imidazoles[1]. It is a versatile synthon in organic chemistry, extensively used for the synthesis of heterocyclic compounds[2].

Application

p-Toluenesulfonylmethyl isocyanide was used:
  • in the synthesis of triplet drugs with the 1,3,5-trioxazatriquinane skeleton[3]
  • as reagent in the preparation of biologically active pyrroles and imidazoles[4][5]
  • as the isonitrile component in a diastereoselective Passerini reaction employing sugar-derived aldehydes[6]

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral

Supplementary Hazards

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

p-Toluenesulfonylmethyl Isocyanide: A Versatile Synthon in Organic Chemistry.
Tandon VK and Rai S.
Journal of Sulfur Chemistry, 24(3), 307-385 (2003)
Kang Wang et al.
Science advances, 5(6), eaaw2953-eaaw2953 (2019-06-20)
Organic solid-state lasers are essential for various photonic applications, yet current-driven lasing remains a great challenge. Charge transfer (CT) complexes formed with p-/n-type organic semiconductors show great potential in electrically pumped lasers, but it is still difficult to achieve population
Il Farmaco (Societa Chimica Italiana : 1989), 48, 209-209 (1993)
Tetrahedron Letters, 48, 5977-5977 (2006)
Medicinal Chemistry Research, 3, 192-192 (1993)

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service