Skip to Content
Merck
All Photos(1)

Key Documents

D80401

Sigma-Aldrich

Dicyclohexyl ketone

98%

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
$75.40

$75.40

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 G
$75.40

About This Item

Linear Formula:
(C6H11)2CO
CAS Number:
Molecular Weight:
194.31
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$75.40

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

98%

form

liquid

refractive index

n20/D 1.485 (lit.)

bp

138 °C/13 mmHg (lit.)

density

0.986 g/mL at 25 °C (lit.)

SMILES string

O=C(C1CCCCC1)C2CCCCC2

InChI

1S/C13H22O/c14-13(11-7-3-1-4-8-11)12-9-5-2-6-10-12/h11-12H,1-10H2

InChI key

TWXWPPKDQOWNSX-UHFFFAOYSA-N

Related Categories

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Matthew M D Roy et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 22(50), 18236-18246 (2016-11-01)
N-Heterocyclic carbene (NHC) complexes of Cd and Hg triflates (OTf) were prepared and their attempted conversion into rare cadmium and mercury hydrides was explored. In contrast to zinc, which forms stable [ZnH]+ complexes with NHCs, the heavier Cd and Hg
Christopher L Rock et al.
Dalton transactions (Cambridge, England : 2003), 48(2), 461-467 (2018-11-30)
The phosphine-substituted α-diimine Ni precursor, (Ph2PPrDI)Ni, has been found to catalyze alkene hydrosilylation in the presence of Ph2SiH2 with turnover frequencies of up to 124 h-1 at 25 °C (990 h-1 at 60 °C). Moreover, the selective hydrosilylation of allylic

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service