Skip to Content
Merck
All Photos(1)

Key Documents

46746

Sigma-Aldrich

Polyglycolic acid

Synonym(s):

PGA, Poly(2-hydroxyacetic acid)

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:

description

inherent viscosity 1.2 dl/g

InChI

1S/C2H4O3/c3-1-2(4)5/h3H,1H2,(H,4,5)

InChI key

AEMRFAOFKBGASW-UHFFFAOYSA-N

Application

medical device

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Huaping Chen et al.
Biomaterials, 34(16), 4159-4172 (2013-03-14)
The absence of safe, efficient, cost-effective, and easily scalable delivery platforms is one of the most significant hurdles and critical issues that limit the bench to bedside translation of oligonucleotides-based therapeutics. Acid-labile materials are of special interest in developing nonviral
M A Tiunin et al.
Eksperimental'naia i klinicheskaia farmakologiia, 76(1), 3-6 (2013-03-07)
Technological parameters for the effective encapsulation of n-(1-phenethyl-4-piperidyl)propionanilide in poly(lactid-co-glycolide) (PLG) nanoparticles have been determined. Depending on the ratio of drug fractions adsorbed on the particle surface and associated with the polymer matrix, n-(1-phenethyl-4-piperidyl)propionanilide (200 microg/kg, i/m) loaded PLG nanospheres
Rodney A Rosalia et al.
Cancer immunology, immunotherapy : CII, 62(7), 1161-1173 (2013-04-25)
Protein antigen (Ag)-based immunotherapies have the advantage to induce T cells with a potentially broad repertoire of specificities. However, soluble protein Ag is generally poorly cross-presented in MHC class I molecules and not efficient in inducing robust cytotoxic CD8(+) T
Steven Lotz et al.
PloS one, 8(2), e56289-e56289 (2013-02-26)
An essential aspect of stem cell culture is the successful maintenance of the undifferentiated state. Many types of stem cells are FGF2 dependent, and pluripotent stem cells are maintained by replacing FGF2-containing media daily, while tissue-specific stem cells are typically
Mitsuhiro Enjo et al.
Plastic and reconstructive surgery, 131(3), 335e-342e (2013-03-01)
Approaches to auricular reconstruction have shown improved outcome when a basic fibroblast growth factor (bFGF) slow-release system and fibrin spraying are combined with biodegradable polymers. More complex, three-dimensional structures, such as those that replicate the human auricle, are often lost

Articles

Interest in utilizing biodegradable polymers for biomedical applications has grown since the 1960s.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service