Skip to Content
Merck
All Photos(3)

Key Documents

124737

Sigma-Aldrich

Glycolic acid

ReagentPlus®, 99%

Synonym(s):

Hydroxyacetic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOCH2COOH
CAS Number:
Molecular Weight:
76.05
Beilstein:
1209322
EC Number:
MDL number:
UNSPSC Code:
12352100
eCl@ss:
39021303
PubChem Substance ID:
NACRES:
NA.21

vapor pressure

0.00093 hPa ( 25 °C)

Quality Level

product line

ReagentPlus®

Assay

99%

form

solid

bp

(169 °C 336 °F at 998 hPa)

mp

75-80 °C (lit.)

solubility

H2O: 50 mg/mL, clear, colorless

density

(1,26 g/cm3 at 20 °C - OECD Test Guideline 109)

functional group

carboxylic acid
hydroxyl

SMILES string

OCC(O)=O

InChI

1S/C2H4O3/c3-1-2(4)5/h3H,1H2,(H,4,5)

InChI key

AEMRFAOFKBGASW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Glycolic acid (Hydroxyacetic acid) is an α-hydroxy acid. Glycolic acid solutions having concentration of 70% and pH range of 0.08 to 2.75 are widely employed as superficial chemical peeling agents. Various oligomers or polymers of lactic and/or glycolic acid (low molecular weight) have been prepared. Glycolic acid can be determined via plant tissue coupled flow injection chemiluminescence biosensors, which can be used both as a plant-tissue based biosensor and chemiluminescence flow sensor.

Application

Glycolic acid (Hydroxyacetic acid) has been used in the preparation of PLGA-PEG-PLGA copolymer (PLGA = poly(lactic/glycolic, PEG = polyethylene glycol).

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany
Redi-Dri is a trademark of Sigma-Aldrich Co. LLC

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

>572.0 °F - (decomposition)

Flash Point(C)

> 300 °C - (decomposition)

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sima Shahabi et al.
TheScientificWorldJournal, 2014, 420616-420616 (2015-01-13)
Gamma irradiation is able to affect various structural and biological properties of biomaterials In this study, a composite of Hap/PLGA-PEG and their ingredients were submitted to gamma irradiation doses of 25 and 50 KGy. Various properties such as molecular weight (GPC)
?Plant Tissue-Based Chemiluminescence Flow Biosensor for Glycolic Acid?
Li B, et al.
Analytical Chemistry, 73(6), 1203-1206 (2001)
"A novel renewable plant tissue-based electrochemiluminescent biosensor for glycolic acid"
Zhu L, et al.
Sensors and Actuators B, Chemical, 98(2), 115-121 (2004)
N Wang et al.
Journal of biomaterials science. Polymer edition, 8(12), 905-917 (1997-01-01)
A series of oligomers or low molecular weight polymers of lactic and/or glycolic acid has been synthesized with different molar ratios of lactic to glycolic acid. These oligomers have been characterized with respect to oligomer composition, molecular weight, intrinsic viscosity
F F Becker et al.
Dermatologic surgery : official publication for American Society for Dermatologic Surgery [et al.], 22(5), 463-465 (1996-05-01)
Seventy percent glycolic acid solutions are being commonly used as superficial chemical peeling agents. The pH of these solutions ranges from 0.08 to 2.75. The histologic effects of these various pH solutions on human skin have not been studied. The

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service