Skip to Content
Merck
All Photos(1)

Key Documents

764590

Sigma-Aldrich

Poly(L-lactide)

average Mn 5,000, PDI ≤1.2

Synonym(s):

PLLA, Polylactide, L-Lactide polymer, PLA, Poly(L-Lactic acid)

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$432.00

$432.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$432.00

About This Item

Linear Formula:
H(C3H4O2)nOCH3
UNSPSC Code:
12162002
NACRES:
NA.23

$432.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

form

solid

optical activity

[α]22/D -147°, c = 0.5% in chloroform

mol wt

average Mn 5,000

degradation timeframe

>3 years

transition temp

Tm 149-154 °C

PDI

≤1.2

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

Poly (L-lactide) is a semi-crystalline polymer which exhibits strong optical rotation and good mechanical properties. It is mostly used in the synthesis of degradable polymers which are used in orthopaedic devices, in dental applications, as scaffolds for autografted new skin, wound covers, anastomose systems and stents. They are biodegradable since the polyester backbone degrades by simple hydrolysis to form non-toxic compounds.[1]

Application

Drug Delivery including coating nanoparticles[2];end-group functionalization[3] ;macroinitiator or block precursor.[4]

Features and Benefits

High initial strength, good strength retention and high amount of crystallinity as much as 70% which in turn is responsible for the very slow degradation rate.[1]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis and thermal properties of novel star-shaped poly (l-lactide)s with starburst PAMAM-OH dendrimer macroinitiator.
Zhao, Y. L., Cai, Q., Jiang, J., Shuai, X. T., Bei, J. Z., Chen, C. F., & Xi, F.
Polymer, 43(22), 5819-5825 (2002)
Chemical synthesis of polylactide and its copolymers for medical applications
Bendix, D.
Polymer Degradation and Stability, 59(1), 129-135 (1998)
Effect of poly (ethylene glycol)-block- poly (L-lactide) on the poly [(R)-3-hydroxybutyrate]/poly (L-lactide) blends
Yoon, J. S., Lee, W. S., Kim, K. S., Chin, I. J., Kim, M. N., & Kim, C.
Eur. Polymer J., 36(2), 435-442 (2000)
Zhang, Wen-Bin; et al.
Macromolecules, 44, 2589-2596 (2011)
Wolf, Florian F.; et al.
Macromolecules, 42, 5622-5628 (2009)

Articles

Professor Aran discusses engineering graphene-based materials through careful functionalization, enabling diverse applications.

Local delivery of bioactive molecules using an implantable device can decrease the amount of drug dose required as well as non-target site toxicities compared to oral or systemic drug administration.

Synthetic aliphatic polyesters dominate resorbable biomaterials in clinical use.

AliAliphatic polyesters, including polylactide and polyglycolide, are biodegradable polymers widely used in medical applications.

See All

Questions

  1. 本製品の形状はsolidとありますが、パウダーでしょうか?それよりも大きなフレーク状でしょうか?

    1 answer
    1. The physical form specification for this product is listed as 'Powder or Solid or Chunk(s)'. The appearance is reported on a lot-specific basis. Please see the link below to review a sample Certificate of Analysis:
      https://www.sigmaaldrich.com/certificates/COFA/76/764590/764590-BULK_______MKCN1974__.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service