Skip to Content
Merck
All Photos(1)

Documents

252476

Sigma-Aldrich

Lactic acid solution

ACS reagent, ≥85%

Synonym(s):

2-Hydroxypropionic acid solution, DL-Lactic acid solution

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH(OH)CO2H
CAS Number:
Molecular Weight:
90.08
Beilstein:
5238667
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.21

grade

ACS reagent

Quality Level

Assay

85.0-90.0%

concentration

≥85%
85.0-90.0% in H2O

impurities

H2SO4, passes test (darkened)

ign. residue

≤0.02%

refractive index

n20/D 1.4251

density

1.209 g/mL at 25 °C

anion traces

chloride (Cl-): ≤0.001%
sulfate (SO42-): ≤0.002%

cation traces

Fe: ≤5 ppm
heavy metals: ≤5 ppm (by ICP)

SMILES string

CC(O)C(O)=O

InChI

1S/C3H6O3/c1-2(4)3(5)6/h2,4H,1H3,(H,5,6)

InChI key

JVTAAEKCZFNVCJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Lactic acid solution has been used in the preparation of poly(lactic acid) (PLA)/lactic acid (LA) blends to enhance the biodegradability rate of PLA.

Other Notes

Contains varying amounts of intermolecular esterification products.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1C

Supplementary Hazards

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Biodegradability of poly (lactic acid)(PLA)/lactic acid (LA) blends using anaerobic digester sludge
Lee JC, et al
Macromolecular Research, 24(8), 741-747 (2016)
A literature review of poly (lactic acid).
Garlotta D.
Journal of Polymers and the Environment, 9(2), 63-84 (2001)
Kwon et al.
Enzyme and microbial technology, 26(2-4), 209-215 (2000-02-26)
Batch fermentation studies were performed to evaluate the potentials of a complex nitrogen source, soybean, as an alternative to yeast extract for the economical production of lactic acid by Lactobacillus rhamnosus. An enzyme-hydrolysate of soybean meal, Soytone, with an adequate
L (+) lactic acid fermentation and its product polymerization.
Narayanan N, et al.
Electronic journal of Biotechnology, 7(2), 167-178 (2004)
Mauro Vasconi et al.
Foods (Basel, Switzerland), 9(5) (2020-05-01)
Processed fish roes are acquiring considerable importance in the modern food market, entering more and more often as an ingredient in food preparation and as caviar substitutes. In this study, we defined quality, traceability and safety issues related to processed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service