Skip to Content
Merck
All Photos(1)

Key Documents

420875

Sigma-Aldrich

LUDOX® AM colloidal silica

30 wt. % suspension in H2O

Synonym(s):

Silica preparation

Sign Into View Organizational & Contract Pricing

Select a Size

1 L
$54.50
4 L
$151.00
18 L
$406.00

$54.50

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 L
$54.50
4 L
$151.00
18 L
$406.00

About This Item

Linear Formula:
SiO2
CAS Number:
Molecular Weight:
60.08
MDL number:
UNSPSC Code:
12352309
PubChem Substance ID:
NACRES:
NA.22

$54.50

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

description

colloidal

Quality Level

form

liquid

concentration

30 wt. % suspension in H2O

surface area

198-250 m2/g

pH

8.6-9.3

density

1.21 g/mL at 25 °C

SMILES string

O=[Si]=O

InChI

1S/O2Si/c1-3-2

InChI key

VYPSYNLAJGMNEJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

contains sodium stabilizing counterion.

LUDOX® AM colloidal silica consists of more condensed silicon structure and it is used in the preparation of polyaniline dispersions.[1][2]

Application

LUDOX® AM colloidal silica can be used as a source of silica for the synthesis of:
  • NaX zeolites by hydrothermal method.[3]
  • Chrysotile-type Mg3Si2O5·(OH)4 nanotubes (MgSi-NTs).[4]

It can also be used as a binder for ceramic slurries.[5]

Legal Information

LUDOX is a registered trademark of W.R. Grace & Co.-Conn.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Skin Sens. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Investigation of the Use of Various Silica Source on NaX Zeolite Properties
Eskandari A, et al.
Journal of Chemical and Petroleum Engineering, 50(2), 1-7 (2017)
Polyaniline dispersions. 6. Stabilization by colloidal silica particles
Stejskal
Macromolecules, 29, 6814-6819 (1996)
Synthesis and catalytic activity of chrysotile-type magnesium silicate nanotubes using various silicate sources.
Lopez-Salinas E, et al.
Microporous and Mesoporous Materials : The Official Journal of the International Zeolite Association, 274(2), 176-182 (2019)
Particle morphology and chemical microstructure of colloidal silica spheres made from alkoxysilanes
Van Blaaderen
Journal of Non-Crystalline Solids, 149, 161-178 (1992)
Influence of the Al2O3 Solid Phase on the Kinetics of Binding Ceramic Moulds.
Kolczyk J, et al.
Archives of Foundry Engineering, 17(4), 91-96 (2017)

Questions

1–3 of 3 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  3. What is the smallest size of the silica particles?

    1 answer
    1. The Ludox products have an average particle size range of 7nm to 22nm. Please see the Ludox product comparison table below:

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service