Skip to Content
Merck
All Photos(1)

Key Documents

420840

Sigma-Aldrich

LUDOX® AS-40 colloidal silica

40 wt. % suspension in H2O

Synonym(s):

Silica preparation

Sign Into View Organizational & Contract Pricing

Select a Size

1 L
$82.10
4 L
$198.00
18 L
$668.00

$82.10

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 L
$82.10
4 L
$198.00
18 L
$668.00

About This Item

Linear Formula:
SiO2
CAS Number:
Molecular Weight:
60.08
MDL number:
UNSPSC Code:
12352309
PubChem Substance ID:
NACRES:
NA.22

$82.10

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

description

colloidal

form

liquid

concentration

40 wt. % suspension in H2O

surface area

~135 m2/g

pH

9.1

density

1.295 g/mL at 25 °C

SMILES string

O=[Si]=O

InChI

1S/O2Si/c1-3-2

InChI key

VYPSYNLAJGMNEJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

LUDOX®  AS-40 colloidal silica is used as a silica source in the preparation of zeolites.[1]

Application

Ludox AS-40 is used as:
  • A filler material in the nanocomposite Nafion films[2]
  • A precursor in the synthesis of micro-mesoporous carbon spheres[3]

Legal Information

LUDOX is a registered trademark of W.R. Grace & Co.-Conn.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

New opportunities in Stober synthesis: preparation of microporous and mesoporous carbon spheres
J Choma, et al.
Journal of Materials Chemistry, 22, 12636-12642 (2012)
Separation of water?acetic acid mixtures by pervaporation using a thin mordenite membrane.
Li G, et al.
Separation and Purification Technology, 32(1-3), 199-206 (2003)
The static aging effect on the seedless synthesis of different ranges FAU-type zeolite Y at various factors
Al-Jubouri, et al.
Iraqi Journal of Chemical and Petroleum Engineering, 20, 7-13 (2019)
Salt enhanced solvent relaxation and particle surface area determination via rapid spin-lattice NMR.
Elliott L N, et al.
Powder Technology, 333, 458-467 (2018)
Interfacial Water Drives Improved Proton Transport in Siliceous Nanocomposite Nafion Thin Films
PJ Goossens, et al.
ChemPhysChem, 19, 538-546 (2018)

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service