Skip to Content
Merck
All Photos(1)

Key Documents

ERMFD100

Silica, colloidal

ERM®, certified reference material

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$461.00

$461.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$461.00

About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

$461.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

grade

certified reference material

Agency

ERM®

manufacturer/tradename

JRC

format

matrix material

Looking for similar products? Visit Product Comparison Guide

Application

Silica, colloidal has been used as a reference standard in the quantitative determination of silica nanoparticles in food using dynamic light scattering inductively coupled plasma mass spectrometry (DLS-ICP-MS).[1]

Analysis Note

For more information please see:
ERMFD100

Legal Information

ERM is a registered trademark of European Commission

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Production of reference materials for the detection and size determination of silica nanoparticles in tomato soup
Grombe R, et al.
Analytical and Bioanalytical Chemistry, 406(16), 3895-3907 (2014)
Margarida Bastos et al.
Biophysical journal, 94(6), 2128-2141 (2007-11-23)
The energetics and partition of two hybrid peptides of cecropin A and melittin (CA(1-8)M(1-18) and CA(1-7)M(2-9)) with liposomes of different composition were studied by time-resolved fluorescence spectroscopy, isothermal titration calorimetry, and surface plasmon resonance. The study was carried out with

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service