Skip to Content
Merck
All Photos(1)

Key Documents

420891

Sigma-Aldrich

LUDOX® CL-X colloidal silica

45 wt. % suspension in H2O

Synonym(s):

Silica preparation

Sign Into View Organizational & Contract Pricing

Select a Size

1 L
$79.60
18 L
$824.00

$79.60

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 L
$79.60
18 L
$824.00

About This Item

Linear Formula:
SiO2
CAS Number:
Molecular Weight:
60.08
MDL number:
UNSPSC Code:
12352309
PubChem Substance ID:
NACRES:
NA.22

$79.60

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

description

colloidal

form

suspension

contains

sodium-stabilizing counterion

concentration

45 wt. % suspension in H2O

surface area

~130 m2/g

impurities

7% ethylene glycol

pH

9.1

density

1.36 g/mL at 25 °C

SMILES string

O=[Si]=O

InChI

1S/O2Si/c1-3-2

InChI key

VYPSYNLAJGMNEJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

LUDOX is a registered trademark of W.R. Grace & Co.-Conn.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jacob Beal et al.
Communications biology, 3(1), 512-512 (2020-09-19)
Optical density (OD) is widely used to estimate the density of cells in liquid culture, but cannot be compared between instruments without a standardized calibration protocol and is challenging to relate to actual cell count. We address this with an
Graham J Edgar et al.
Nature, 506(7487), 216-220 (2014-02-07)
In line with global targets agreed under the Convention on Biological Diversity, the number of marine protected areas (MPAs) is increasing rapidly, yet socio-economic benefits generated by MPAs remain difficult to predict and under debate. MPAs often fail to reach
Yaswanth Kuthati et al.
Journal of nanoscience and nanotechnology, 13(4), 2399-2430 (2013-06-15)
The advent of Nanotechnology has paved a way for improved disease treatment strategies, the most noteworthy being the mesoporous silica nanoparticles (MSNs) which have gained much recent attention in the field of cancer therapy and its diagnosis. The flaws of
David M Brown et al.
Toxicology letters, 224(1), 147-156 (2014-01-28)
We investigated the effects of silica particles and nanoparticles (NPs) (50 nm and 200 nm) with a neutral and positively charged surface when dispersed in saline, bovine serum albumin (BSA) or lung lining fluid (LLF) 24 h post instillation into
Andrea F G Gargano et al.
Journal of chromatography. A, 1351, 70-81 (2014-06-05)
Mucin glycoproteins belong to a class of high molecular weight, heavy glycosylated, proteins that together with water, salts and lipids constitute mucous secretions. Particular disease states (e.g. obstructive chronic bronchitis and ovarian tumor) are known to modify the composition and

Questions

  1. What is the smallest size of the silica particles?

    1 answer
    1. The Ludox products have an average particle size range of 7nm to 22nm. Please see the Ludox product comparison table below:

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service