Skip to Content
Merck
All Photos(2)

Key Documents

372595

Sigma-Aldrich

Lead(II) sulfide

99.9% trace metals basis

Synonym(s):

Lead monosulfide, Lead sulfide, Natural lead sulfide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
PbS
CAS Number:
Molecular Weight:
239.27
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
Assay:
99.9% trace metals basis
Pricing and availability is not currently available.

Quality Level

Assay

99.9% trace metals basis

density

7.5 g/mL at 25 °C (lit.)

SMILES string

[PbH2]=S

InChI

1S/Pb.S

InChI key

XCAUINMIESBTBL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

PbS can be precipitated from a solution of lead (II) salt and hydrogen sulfide.[1]

Application

PbS may be used for telecommunications. 2

Packaging

Packaged in poly bottles

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Repr. 1A - STOT RE 2

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

John Dainith
Chemistry (Weinheim An Der Bergstrasse, Germany) null
Jeffrey Yan et al.
Analytical chemistry, 84(14), 6122-6127 (2012-06-13)
Sulfur anions and their derivatives have long been recognized for their high selectivity and reactivity toward Pb(2+) ions and formation of highly absorptive yet water-insoluble compounds with both acid and base media. This phenomenon has been used for qualitative analysis
Illan J Kramer et al.
Advanced materials (Deerfield Beach, Fla.), 24(17), 2315-2319 (2012-04-03)
A bulk heterojunction of ordered titania nanopillars and PbS colloidal quantum dots is developed. By using a pre-patterned template, an ordered titania nanopillar matrix with nearest neighbours 275 nm apart and height of 300 nm is fabricated and subsequently filled
Zhenhua Sun et al.
Advanced materials (Deerfield Beach, Fla.), 24(43), 5878-5883 (2012-09-01)
Infrared photodetectors based on single-layer CVD-grown graphene and PbS quantum dots, which are fabricated by solution processing, show ultrahigh responsivities of up to 10(7) A/W under infrared light illumination. The devices fabricated on flexible plastic substrates have excellent bending stability.
Somobrata Acharya et al.
Nano letters, 13(2), 409-415 (2013-01-10)
Two-dimensional (2D) sheets are currently in the spotlight of nanotechnology owing to high-performance device fabrication possibilities. Building a free-standing quantum sheet with controlled morphology is challenging when large planar geometry and ultranarrow thickness are simultaneously concerned. Coalescence of nanowires into

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service