Skip to Content
Merck
All Photos(2)

Key Documents

308285

Sigma-Aldrich

Indium(III) bromide

99%

Synonym(s):

Indium tribromide

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
$183.00

$183.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 G
$183.00

About This Item

Linear Formula:
InBr3
CAS Number:
Molecular Weight:
354.53
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
form:
powder and chunks

$183.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

vapor density

>1 (vs air)

Quality Level

Assay

99%

form

powder and chunks

reaction suitability

reagent type: catalyst
core: indium

bp

656 °C (lit.)

mp

436 °C (lit.)

density

4.96 g/mL at 25 °C (lit.)

SMILES string

Br[In](Br)Br

InChI

1S/3BrH.In/h3*1H;/q;;;+3/p-3

InChI key

JKNHZOAONLKYQL-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

Application


  • Investigation of the Solution Chemistry of Hybrid Organic–Inorganic Indium Halides for New Material Discovery: The study examines the indium(III) bromide system, MA–In–Br, and the formation of new indium(III) halides (Zhang et al., 2022).

  • Indium (III)‐Catalyzed Haloalkynylation Reaction of Alkynes: This research explores the use of indium(III) bromide as a catalyst in haloalkynylation reactions (Daniels et al., 2024).

  • Zero-Dimensional Hybrid Organic–Inorganic Indium Bromide with Blue Emission: The study reports on the synthesis and properties of a new indium bromide compound exhibiting blue emission (Fattal et al., 2021).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Karavadhi Surendra et al.
Journal of the American Chemical Society, 133(25), 9724-9726 (2011-06-08)
InI(3) and InBr(3) have been found to be effective catalysts for the π activation of C≡C bonds to initiate the conversion of chiral propargylic alcohols or silyl ethers to polycyclic products in excellent yields and with high stereoselectivity. The method

Articles

Colloidal quantum dots (CQDs) are semiconducting crystals of only a few nanometers (ca. 2–12 nm) coated with ligand/surfactant molecules to help prevent agglomeration.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service